Wheat Germ Agglutinin (WGA)
Invitrogen™

Wheat Germ Agglutinin (WGA)

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) and Alexa Fluor, Alexa Fluor Plus, and other dyes. Fluorescent WGA conjugates bind to carbohydrates and are used for various cell biology applications such as plasma membrane labeling and cell painting assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugate
W6748Oregon Green 488
W11263Alexa Fluor 350
W7024Alexa Fluor 350, Oregon Green 488, Tetramethylrhodamine, Texas Red
W11261Alexa Fluor 488
W32464Alexa Fluor 555
W11262Alexa Fluor 594
W21404Alexa Fluor 633
W32466Alexa Fluor 647
W32465Alexa Fluor 680
W56132Alexa Fluor Plus 405
W56133Alexa Fluor Plus 568
W56134Alexa Fluor Plus 770
W834Fluorescein
W849Tetramethylrhodamine
W21405Texas Red-X
Catalog number W6748
Price (USD)
355.88
Special offer
Online exclusive
Ends: 31-Dec-2026
434.00
Save 78.12
5 mg
Conjugate:
Oregon Green 488
Price (USD)
355.88
Special offer
Online exclusive
Ends: 31-Dec-2026
434.00
Save 78.12
5 mg

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates. These lectins can bind to carbohydrates and are available conjugated to Alexa Fluor™, Alexa Fluor™ Plus, and other fluorescent dyes. Fluorescent wheat germ agglutinin conjugates are valuable tools in molecular and cell biology research, enabling researchers to label the plasma membrane in fluorescence imaging and cell painting assays, and study and analyze glycosylation patterns and glycan-mediated processes in cells and tissues.

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) with Alexa Fluor, Alexa Fluor Plus, and other dyes. WGA is a cell impermeant stain that selectively binds to N‐acetylglucosamine and N‐acetylneuraminic acid (sialic acid) residues, which are often found on cell membranes. Fluorescent WGA conjugates provide selective labeling of the plasma membrane with minimal background in many cell types that is retained after formaldehyde fixation and permeabilization with Triton X-100.

These fluorescent lectin conjugates can also be used to label fixed cells; however, to avoid labeling intracellular components, formaldehyde-fixed cells should not be permeabilized before labeling. Fluorescent WGA conjugates are used as plasma membrane stains along with other cellular markers in cell painting assays to provide a phenotypic readout of cell health or cytotoxicity. The Wheat Germ Agglutinin, Alexa Fluor 555 Conjugate is included in the Image-iT Cell Painting Kit (Cat. Nos. I65000 and I65500).

WGA conjugates are also used as retrograde tracers for neuronal tracing experiments and have been shown to cross synapses. These fluorescent lectins are applicable in microbiology studies to label yeast bud scars, the cell membrane of gram-positive but not gram-negative bacteria, and chitin in fungal cell walls. In solution, WGA exists as a heterodimer with a molecular weight of approximately 38,000 Daltons and is normally cationic under physiological conditions. Our WGA conjugates have been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), flow cytometry (FC), and a wide range of chemical, biochemical and immunological assays.

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates with options covering the entire wavelength range. The Wheat Germ Agglutinin Sampler Kit (Cat. No. W7024) includes introductory samples of four fluorescent WGAs: Alexa Fluor 350, Oregon Green 488, tetramethylrhodamine, and Texas Red-X conjugates. The red-fluorescent Alexa Fluor 594 wheat germ agglutinin (WGA) conjugate is also included in the Image-iT LIVE Plasma Membrane and Nuclear Labeling Kit (Cat. No. I34406). The Texas Red-X WGA conjugate can be purchased in the ViaGram Red+ Bacterial Gram Stain and Viability Kit (Cat. No. V7023) to differentiate gram-positive and gram-negative bacteria.

Wheat germ agglutinin (WGA) conjugates fluorescence excitation/emission
W11261 WGA, Alexa Fluor 488 conjugate: 495 nm/519 nm
W11263 WGA, Alexa Fluor 350 conjugate: 346 nm/442 nm
W32464 WGA, Alexa Fluor 555 conjugate: 555 nm/580 nm
W11262 WGA, Alexa Fluor 594 conjugate: 590 nm/617 nm
W21404 WGA, Alexa Fluor 633 conjugate: 632 nm/647 nm
W32466 WGA, Alexa Fluor 647 conjugate: 650 nm/665 nm
W32465 WGA, Alexa Fluor 680 conjugate: 679 nm/702 nm
W56132 WGA, Alexa Fluor Plus 405 conjugate: 408 nm/450 nm
W56133 WGA, Alexa Fluor Plus 568 conjugate: 562 nm/583 nm
W56134 WGA, Alexa Fluor Plus 770 conjugate: 770 nm/797 nm
W834 WGA, fluorescein conjugate: 494 nm/518 nm
W6748 WGA, Oregon Green 488 conjugate: 496 nm/524 nm
W849 WGA, tetramethylrhodamine conjugate: 555 nm/580 nm
W21405 WGA, Texas Red-X conjugate: 595 nm/615 nm

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Common NameWheat Germ Agglutinin, Oregon Green™ 488 Conjugate
Expression SystemWheat germ
Ligand TypeN-acetylglucosamine and N-acetylneuraminic acid (sialic acid) residues
Product LineOregon Green
ProteinFluorescent lectins
Protein FamilyLectins
Protein FormHeterodimer
Protein SubtypeAgglutinin
Protein TagNone
Purity or Quality GradeSee Certificate of Analysis
Quantity5 mg
Shipping ConditionRoom Temperature
SourceWheat germ
ConjugateOregon Green 488
For Use With (Application)Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Cell Painting
FormLyophilized
RecombinantNative
SpeciesWheat
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

My cells are very sensitive and need to be kept in media as much as possible. Is it possible to label the plasma membrane with fluorescent wheat germ agglutinin (WGA) in media instead of buffer?

Yes. Although labeling in buffer (such as Hank's Buffered Saline Solution) is slightly better for brightness and lower non-cell background, media can be used. Do a concentration range to dertermine optimal conditions, since the WGA may potentially bind media components to some extent, slightly decreasing your specific labeling intensity.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to label the plasma membrane of my cells, but there are several dyes to choose from. Which one should I use?

For live-cell imaging, the CellVue and CellMask Plasma Membrane Stains are the most uniform and the slowest to be endocytosed. However, they are not the best choice if you wish to fix and permeabilize your cells, such as for antibody labeling. Wheat germ agglutinin (WGA) conjugates are also able to label live cells, or can label already formaldehyde-fixed cells. They can survive subsequent permeabilization with detergents, such as Triton X-100. If cells are already permeabilized, WGA will label internal structures as well. Thus, only an antibody against a plasma membrane protein can be used if cells are already permeabilized. Lipophilic cyanine dyes, such as DiI, will label all cell membranes in live cells, not just plasma membranes, if left on live cells for extended periods. Following page will help you choose (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-structure/plasma-membrane.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (20)

Citations & References
Abstract
A flow-cytometric gram-staining technique for milk-associated bacteria.
Authors:Holm C, Jespersen L
Journal:Appl Environ Microbiol
PubMed ID:12732558
'A Gram-staining technique combining staining with two fluorescent stains, Oregon Green-conjugated wheat germ agglutinin (WGA) and hexidium iodide (HI) followed by flow-cytometric detection is described. WGA stains gram-positive bacteria while HI binds to the DNA of all bacteria after permeabilization by EDTA and incubation at 50 degrees C for 15 ... More
Loss of abaxial leaf epicuticular wax in Medicago truncatula irg1/palm1 mutants results in reduced spore differentiation of anthracnose and nonhost rust pathogens.
Authors:Uppalapati SR, Ishiga Y, Doraiswamy V, Bedair M, Mittal S, Chen J, Nakashima J, Tang Y, Tadege M, Ratet P, Chen R, Schultheiss H, Mysore KS,
Journal:Plant Cell
PubMed ID:22294617
'To identify genes that confer nonhost resistance to biotrophic fungal pathogens, we did a forward-genetics screen using Medicago truncatula Tnt1 retrotransposon insertion lines. From this screen, we identified an inhibitor of rust germ tube differentation1 (irg1) mutant that failed to promote preinfection structure differentiation of two rust pathogens, Phakopsora pachyrhizi ... More
Development of a quantitative, cell-based, high-content screening assay for epidermal growth factor receptor modulators.
Authors:Wang J, Xie X,
Journal:Acta Pharmacol Sin
PubMed ID:17883960
'AIM: To develop a robust, cell-based, high-content screening (HCS) assay based on receptor internalization for the identification of novel modulators of the epidermal growth factor receptor (EGFR). METHODS: Agonist-induced receptor internalization is part of the signaling cascade of EGFR. Fluorescent-tagged epidermal growth factor (EGF) was used to visualize the internalized ... More
Sympathetic alpha(2)-adrenoceptors prevent cardiac hypertrophy and fibrosis in mice at baseline but not after chronic pressure overload.
Authors:Gilsbach R, Schneider J, Lother A, Schickinger S, Leemhuis J, Hein L,
Journal:Cardiovasc Res
PubMed ID:20083574
'alpha(2)-Adrenoceptors modulate cardiovascular function by vasoconstriction or dilatation, by central inhibition of sympathetic activity, or by feedback inhibition of norepinephrine release from sympathetic neurons. Despite detailed knowledge about subtype-specific functions of alpha(2)-receptors, the relative contributions of sympathetic vs. non-sympathetic receptors involved in these cardiovascular effects have not been identified. The ... More
Murine hyaluronidase 2 deficiency results in extracellular hyaluronan accumulation and severe cardiopulmonary dysfunction.
Authors:Chowdhury B, Hemming R, Hombach-Klonisch S, Flamion B, Triggs-Raine B,
Journal:J Biol Chem
PubMed ID:23172227
Hyaluronidase (HYAL) 2 is a membrane-anchored protein that is proposed to hydrolyze hyaluronan (HA) to smaller fragments that are internalized for breakdown. Initial studies of a Hyal2 knock-out (KO) mouse revealed a mild phenotype with high serum HA, supporting a role for HYAL2 in HA breakdown. We now describe a ... More