Wheat Germ Agglutinin (WGA)
Invitrogen™

Wheat Germ Agglutinin (WGA)

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) and Alexa Fluor, Alexa Fluor Plus, and other dyes. Fluorescent WGA conjugates bind to carbohydrates and are used for various cell biology applications such as plasma membrane labeling and cell painting assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugate
W32464Alexa Fluor 555
W11263Alexa Fluor 350
W7024Alexa Fluor 350, Oregon Green 488, Tetramethylrhodamine, Texas Red
W11261Alexa Fluor 488
W11262Alexa Fluor 594
W21404Alexa Fluor 633
W32466Alexa Fluor 647
W32465Alexa Fluor 680
W56132Alexa Fluor Plus 405
W56133Alexa Fluor Plus 568
W56134Alexa Fluor Plus 770
W834Fluorescein
W6748Oregon Green 488
W849Tetramethylrhodamine
W21405Texas Red-X
Catalog number W32464
Price (USD)
411.64
Special offer
Online exclusive
Ends: 31-Dec-2026
502.00
Save 90.36
5 mg
Conjugate:
Alexa Fluor 555
Price (USD)
411.64
Special offer
Online exclusive
Ends: 31-Dec-2026
502.00
Save 90.36
5 mg

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates. These lectins can bind to carbohydrates and are available conjugated to Alexa Fluor™, Alexa Fluor™ Plus, and other fluorescent dyes. Fluorescent wheat germ agglutinin conjugates are valuable tools in molecular and cell biology research, enabling researchers to label the plasma membrane in fluorescence imaging and cell painting assays, and study and analyze glycosylation patterns and glycan-mediated processes in cells and tissues.

Thermo Fisher Scientific offers bright conjugates of wheat germ agglutinin (WGA) with Alexa Fluor, Alexa Fluor Plus, and other dyes. WGA is a cell impermeant stain that selectively binds to N‐acetylglucosamine and N‐acetylneuraminic acid (sialic acid) residues, which are often found on cell membranes. Fluorescent WGA conjugates provide selective labeling of the plasma membrane with minimal background in many cell types that is retained after formaldehyde fixation and permeabilization with Triton X-100.

These fluorescent lectin conjugates can also be used to label fixed cells; however, to avoid labeling intracellular components, formaldehyde-fixed cells should not be permeabilized before labeling. Fluorescent WGA conjugates are used as plasma membrane stains along with other cellular markers in cell painting assays to provide a phenotypic readout of cell health or cytotoxicity. The Wheat Germ Agglutinin, Alexa Fluor 555 Conjugate is included in the Image-iT Cell Painting Kit (Cat. Nos. I65000 and I65500).

WGA conjugates are also used as retrograde tracers for neuronal tracing experiments and have been shown to cross synapses. These fluorescent lectins are applicable in microbiology studies to label yeast bud scars, the cell membrane of gram-positive but not gram-negative bacteria, and chitin in fungal cell walls. In solution, WGA exists as a heterodimer with a molecular weight of approximately 38,000 Daltons and is normally cationic under physiological conditions. Our WGA conjugates have been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), flow cytometry (FC), and a wide range of chemical, biochemical and immunological assays.

Thermo Fisher Scientific offers a broad selection of fluorescent wheat germ agglutinin conjugates with options covering the entire wavelength range. The Wheat Germ Agglutinin Sampler Kit (Cat. No. W7024) includes introductory samples of four fluorescent WGAs: Alexa Fluor 350, Oregon Green 488, tetramethylrhodamine, and Texas Red-X conjugates. The red-fluorescent Alexa Fluor 594 wheat germ agglutinin (WGA) conjugate is also included in the Image-iT LIVE Plasma Membrane and Nuclear Labeling Kit (Cat. No. I34406). The Texas Red-X WGA conjugate can be purchased in the ViaGram Red+ Bacterial Gram Stain and Viability Kit (Cat. No. V7023) to differentiate gram-positive and gram-negative bacteria.

Wheat germ agglutinin (WGA) conjugates fluorescence excitation/emission
W11261 WGA, Alexa Fluor 488 conjugate: 495 nm/519 nm
W11263 WGA, Alexa Fluor 350 conjugate: 346 nm/442 nm
W32464 WGA, Alexa Fluor 555 conjugate: 555 nm/580 nm
W11262 WGA, Alexa Fluor 594 conjugate: 590 nm/617 nm
W21404 WGA, Alexa Fluor 633 conjugate: 632 nm/647 nm
W32466 WGA, Alexa Fluor 647 conjugate: 650 nm/665 nm
W32465 WGA, Alexa Fluor 680 conjugate: 679 nm/702 nm
W56132 WGA, Alexa Fluor Plus 405 conjugate: 408 nm/450 nm
W56133 WGA, Alexa Fluor Plus 568 conjugate: 562 nm/583 nm
W56134 WGA, Alexa Fluor Plus 770 conjugate: 770 nm/797 nm
W834 WGA, fluorescein conjugate: 494 nm/518 nm
W6748 WGA, Oregon Green 488 conjugate: 496 nm/524 nm
W849 WGA, tetramethylrhodamine conjugate: 555 nm/580 nm
W21405 WGA, Texas Red-X conjugate: 595 nm/615 nm

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Common NameWheat Germ Agglutinin, Alexa Fluor™ 555 Conjugate
Expression SystemWheat germ
Ligand TypeN-acetylglucosamine and N-acetylneuraminic acid (sialic acid) residues
Product LineAlexa Fluor
ProteinFluorescent lectins
Protein FamilyLectins
Protein FormHeterodimer
Protein SubtypeAgglutinin
Protein TagNone
Purity or Quality GradeSee Certificate of Analysis
Quantity5 mg
Shipping ConditionRoom Temperature
SourceWheat germ
ConjugateAlexa Fluor 555
For Use With (Application)Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Cell Painting
FormLyophilized
RecombinantNative
SpeciesWheat
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

My cells are very sensitive and need to be kept in media as much as possible. Is it possible to label the plasma membrane with fluorescent wheat germ agglutinin (WGA) in media instead of buffer?

Yes. Although labeling in buffer (such as Hank's Buffered Saline Solution) is slightly better for brightness and lower non-cell background, media can be used. Do a concentration range to dertermine optimal conditions, since the WGA may potentially bind media components to some extent, slightly decreasing your specific labeling intensity.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use CellMask Plasma membrane stains or Alexa Fluor dye labeled wheat germ agglutinin to label the plasma membrane of my paraffin sections?

No. For paraffin sections, there are few options due to the delipidation of the membranes by solvents used in the deparaffinization steps. The only good option is to use an antibody against a plasma membrane protein.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to label Golgi bodies in my fixed cells. Is a dye-conjugated lectin like wheat germ agglutinin or concanavalin A a good choice?

No. Those lectins have been shown to label Golgi in fixed and permeabilized cultured cells, but the selectivity is cell type dependent. Usually you wind up with other structures labeling as well, such as endoplasmic reticulum. The only guaranteed way to specifically label Golgi in already fixed cells is to use an antibody for a Golgi-specific antigen.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What should I consider when working with Wheat Germ Agglutinin, Alexa Fluor 555 Conjugate (Cat. No. W32464), so as to selectively label plasma membrane rather than intra-cellular structures?

If the cells are permeabilized, WGA will label intracellular structures (i.e., the Golgi bodies, ER, vesicles etc., varying by cell type) as well as the plasma membrane. If you want to label the plasma membrane only, then you must either label live cells or label after fixation with an alcohol-free formaldehyde prior to any permeabilization. Please note that other fixatives, such as alcohol or acetone, will permeabilize the membrane and result in intracellular labeling. Other considerations are as follows: when you selectively label the plasma membrane, the focal depth is an issue for cultured cells, where with a widefield microscopy you will be imaging through the entire cell. Therefore, if the label is only at the top or bottom of the cell on the plasma membrane, it will falsely appear as if it is internalized, because the focal depth is not narrow enough. Using a confocal system with a small pinhole and a high enough magnification objective can sometimes overcome this, or by having the cells grow confluent prior to labeling, which gives them more three-dimensionality.

Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.

I injected a fluorescent tracer, but cannot detect it after tissue is fixed and sectioned. What am I doing wrong?

Confirm that the tracer you are using crosslinks to proteins or has a primary amine for fixation-either a hydrazide, lysine fixable dextran, or a protein conjugate.
Use aldehyde-based fixatives to cross link the amines on the tracer.
Inject a larger amount or higher concentration of the tracer. Tracers are generally injected at 1-20% concentrations (10 mg/mL or higher).
Confirm that you are using the correct fluorescent filter for detection. You can perform a spot test by pipetting a small amount of the undiluted stock solution of the tracer onto a slide, then view under the filter you are using on your microscope. This will confirm if the tracer fluorescence can be detected and the fluorescent microscope filter is working properly.
Review tissue fixation and handling procedures to confirm if any reagents or processing procedures could be affecting the tracer.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (24)

Citations & References
Abstract
Dysmorphic photoreceptors in a P23H mutant rhodopsin model of retinitis pigmentosa are metabolically active and capable of regenerating to reverse retinal degeneration.
Authors:Lee DC, Vazquez-Chona FR, Ferrell WD, Tam BM, Jones BW, Marc RE, Moritz OL,
Journal:J Neurosci
PubMed ID:22323724
'This study evaluated the capacity of Xenopus laevis retina to regenerate photoreceptor cells after cyclic light-mediated acute rod photoreceptor degeneration in a transgenic P23H mutant rhodopsin model of retinits pigmentosa. After discontinuation of cyclic light exposure, we monitored histologic progression of retinal regeneration over a 3 week recovery period. To ... More
Phosphoglycerate mutase affects Stenotrophomonas maltophilia attachment to biotic and abiotic surfaces.
Authors:Ramos-Hegazy L, Chakravarty S, Anderson GG
Journal:
PubMed ID:31430538
Stenotrophomonas maltophilia biofilm formation is of increasing medical concern, particularly for lung infections. However, the molecular mechanisms facilitating the biofilm lifestyle in S. maltophilia are poorly understood. We generated and screened a transposon mutant library for mutations that lead to altered biofilm formation compared to wild type. One of these mutations, ... More
Axon sorting within the spinal cord marginal zone via Robo-mediated inhibition of N-cadherin controls spinocerebellar tract formation.
Authors:Sakai N, Insolera R, Sillitoe RV, Shi SH, Kaprielian Z,
Journal:J Neurosci
PubMed ID:23115176
The axons of spinal projection neurons transmit sensory information to the brain by ascending within highly organized longitudinal tracts. However, the molecular mechanisms that control the sorting of these axons within the spinal cord and their directed growth to poorly defined targets are not understood. Here, we show that an ... More
The transmembrane domain of the severe acute respiratory syndrome coronavirus ORF7b protein is necessary and sufficient for its retention in the Golgi complex.
Authors:Schaecher SR, Diamond MS, Pekosz A,
Journal:J Virol
PubMed ID:18632859
The severe acute respiratory syndrome coronavirus (SARS-CoV) ORF7b (also called 7b) protein is an integral membrane protein that is translated from a bicistronic open reading frame encoded within subgenomic RNA 7. When expressed independently or during virus infection, ORF7b accumulates in the Golgi compartment, colocalizing with both cis- and trans-Golgi ... More
A novel celecoxib analog UTX-121 inhibits HT1080 cell invasion by modulating membrane-type 1 matrix metalloproteinase.
Authors:Yamahana H, Takino T, Endo Y, Yamada H, Suzuki T, Uto Y
Journal:Biochem Biophys Res Commun
PubMed ID:31629465
'We designed and synthesized a celecoxib derivative UTX-121 to enhance its anti-tumor activity. Similar to celecoxib, this compound could also inhibit matrix metalloproteinase (MMP)-9 activity. In addition, UTX-121 suppressed membrane-type 1 MMP (MT1-MMP)-mediated pro-MMP-2 activation by disturbing the cell surface expression of MT1-MMP. UTX-121 also impeded the glycosylation of cell ... More