Search
Amplification of the No Template Control due to reagent contamination
If you get an amplification product in your no template control (NTC), you may have one of these problems:
- Contamination of your reactions by DNA
Reaction contamination can be separated into two types (select one):
- Primer dimer formation (Applied Biosystems™ SYBR™ Green chemistry only)
In this situation, NTC contamination randomly occurs when loading the DNA templates into the PCR plate. If the contamination is due to plate loading, the NTC will show amplification in some or all of the NTCs at varying CT values; see figure below.
NTC contamination
Solution:
One or more reagents are contaminated. If this is the case, amplification of the NTC replicates should be close, as shown below, because the same amount of template was loaded onto the PCR plate.
Amplification as a result of reagent contamination
All reagents used in real-time PCR reactions (i.e., master mix, water, and primers/probes) can become contaminated with template. This can be due to a lack of GLP (good laboratory practice) resulting in random template contaminations. In addition, if you perform pre-PCR and post-PCR procedures in the same area, the PCR product carryover from a previous reaction can contaminate a new PCR plate and cause positive results.
Solution:
When Applied Biosystems™ SYBR™ Green dye is used in real-time PCR, any primer dimers will cause higher background, and may lead to a generation of CT<40 for NTC samples. The primer dimer is not usually detected unless a dissociation profile is generated. Because the background is higher, large amounts of primer dimers may alter the CT of your experimental samples and change the expression level interpretation.
![]() | A SYBR Green amplification plot prior to the dissociation curve. In this view the presence of primer dimers cannot be determined. | |
![]() | A dissociation curve following a SYBR Green real-time reaction. The presence of primer dimers can be easily identified in the dissociation curve by the additional peak(s). The primer dimer peaks are usually seen at low melting temperatures. |
Solution:
| Reverse primer (nM) | Forward primer (nM) | ||
|---|---|---|---|
| 100 | 200 | 400 | |
| 100 | 100/100 | 200/100 | 400/100 |
| 200 | 100/200 | 200/200 | 400/200 |
| 400 | 100/400 | 200/400 | 400/400 |
For Research Use Only. Not for use in diagnostic procedures.