Low DNA Binding Snap Cap Microcentrifuge Tube
On this page:
Lower carbon footprint while maintaining quality and performance
Thermo Scientific Snap Cap Microcentrifuge Tubes, Sustain Series, are an indispensable tool in every laboratory setting, designed with versatility and efficiency to help ensure optimal performance in sample preparation, product transfer, and short-term storage. Notably, these tubes are low DNA binding, helping ensure minimal sample loss, and free from PCR inhibitors, UV leachables, and human DNA, making them excellent for sensitive applications. Furthermore, they are more sustainable – with biobased resin and International Sustainability and Carbon (ISCC) PLUS certification, underscoring their greener design.
DNA Integrity
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to help minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures. Enabling that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays. Overall, the use of low DNA binding microcentrifuge tubes is essential for maintaining the integrity and consistency of DNA samples, helping lead to more accurate and reproducible results in molecular biology research.
Specifications
Data/Description
Low DNA binding
PCR efficiency > 75% after storing 100, 10, 1 copies of gDNA at -80degC for 24 hours.
Human DNA free
hDNA <.03pg/uL
PCR inhibitor free
PCR run down to 0.001 ng/uL gDNA using water incubated at 95degC for 1 hour.
UV leachable free
UV absorbance of water less than 0.02 au (220 and 230 nm) and less than 0.01 au (260 and 280 nm) after 16-18 hour incubation at 65degC.
More Sustainable Solution
More Sustainable Alternative
The Thermo Scientific DNA Low Binding Snap Cap Microcentrifuge Tubes, Sustain Series, offers a greener solution for lab researchers in biotech, pharma, and forensics. Made from ISCC PLUS certified biobased polypropylene derived from second-generation waste and residue oils, these tubes help reduce greenhouse gas emissions—3.43 kg CO2e per kg of biobased polypropylene. By choosing these more sustainable tubes, customers can reduce their carbon footprint, support Scope 3 emission reduction targets, all while maintaining the same performance standards as fossil fuel-based alternatives.
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.