Search
You may need to be an expert in your field, but you shouldn't need to be an expert in dPCR assay design. That's why we've programmed over 40 years of Applied Biosystems assay design experience into the proprietary algorithms powering our new Absolute Q dPCR Custom Assay Design Tool. With this tool, you have years of design expertise at your fingertips to help generate custom probe-based digital PCR (dPCR) assays for research with confidence and ease.
If you know your target, it’s as easy as 1, 2, 3:
Ready to get started with your dPCR experiment, but need support designing custom assays for your targets of interest? With our custom dPCR assay tool you don’t have to be an expert. Let our decades of expertise in PCR assay design ease the process so that you can focus on what matters.
Absolute Q dPCR assays are based on the same gold-standard TaqMan chemistry used in real-time PCR and cited in more than 200K research publications. The assays feature a proprietary probe with a minor groove binder (MGB) moiety at the 3’ end that increases the melting temperature (Tm) of the probe. This means MGB probes can be significantly shorter than traditional approaches, providing high-resolution sequence discrimination and flexibility to accommodate more targets in a single reaction.
Contact us and let your local representative help find the best solution for your custom multiplex dPCR assay design needs.
For Research Use Only. Not for use in diagnostic procedures.