Search
Select from a variety of accessories that offer measurement flexibility for teaching laboratories, research needs, and routine industrial applications. Choose single- and multi-cell sample holders, test tube accessories, temperature control units, sipper systems, fiber optic probes, interface cables, replacement lamps, standards, and more. Browse all spectrophotometer accessories, see accessory compatibility with Thermo Scientific Evolution, GENESYS, or SPECTRONIC 200 spectrophotometers in the table, or scroll down to see descriptions below.
Evolution |
GENESYS |
SPECTRONIC 200 |
|
General accessories |
|||
X |
|
|
|
Liquid sampling accessories |
|
|
|
| Evolution Fiber Optic Probes (b) | X |
|
|
X |
|
|
|
|
X |
|
|
X |
|
|
|
Evolution Smart Thermostatted Linear 8-Cell Changer Accessory |
X |
|
|
Evolution UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer |
X |
|
|
|
X |
|
|
X |
|
|
|
Sipper Systems for GENESYS 40–180 & BioMate 160 Spectrophotometers |
|
X |
|
X |
X |
X |
|
X |
|
|
|
Solid sampling accessories |
|
|
|
X |
|
|
|
X |
|
|
|
Validation tools and calibration standards |
|
|
|
X |
|
|
|
| Stray Light Standards (f) | X |
X |
|
X |
|
|
|
X |
X |
|
|
| SPECTRONIC Spectrophotometer Standards (l) |
|
X |
X |
| Toluene in Hexane Solution Standard (m) | X |
X |
|
| Validation Tools for UV-Visible Spectrophotometers | X |
X |
X |
Additional accessories |
|
|
|
Single-cell Sample Holders for GENESYS and BioMate 160 Spectrophotometers (d) |
|
X |
|
| SPECTRONIC 200 Spectrophotometer Accessories (e) |
|
|
X |
| SPECTRONIC Thermal Printer for SPECTRONIC 200 (k) |
|
|
X |
| Evolution Peltier Thermostatting Accessories (n) | X |
|
|
Product information
a: UV-Vis spectrophotometer calibration validation carousels (CVC) - Thermo Scientific Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money. The CVC provides certified reference materials in a fully automated configuration for hands-free performance verification of your Thermo Scientific spectrophotometer.
b: Evolution Fiber Optic Probes for UV-Vis spectrophotometers - Preserve sample integrity while minimizing sample carryover. Thermo Scientific Evolution Fiber Optic Probes let you bring the spectrophotometer to the sample. Choose the VERSA probe for measurements in larger containers or solutions that attack stainless steel. Choose the Micro-Probe for measurement of small volumes or in small containers. Both are designed for use in small containers such as PCR and centrifuge tubes commonly used in biological applications.
c: Evolution Mercury Lamp accessories for UV-Vis spectrophotometers - Thermo Scientific Mercury Lamp accessories provide instrument calibration and automated performance testing for the Evolution Series UV-Visible spectrophotometers. Test performance of wavelength accuracy, wavelength repeatability and bandwidth accuracy using the extremely narrow emission lines of mercury. These emission lines are fundamental, primary standards that require no certification or recalibration, saving you time and money.
d: Single-cell sample holders for GENESYS and BioMate 160 Spectrophotometers - Make the most of every sample. Accessories for GENESYS UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurement requirements. Cable-free, snap-in design allows for easy exchange of accessories.
e: SPECTRONIC 200 Spectrophotometer accessories - Accessories for SPECTRONIC 200 Spectrophotometer
f: Stray light standards for UV-Vis spectrophotometers - Thermo Scientific NIST traceable standards verify spectrophotometer stray light specification.
g: Potassium dichromate solution standard for UV-Vis spectrophotometers - Thermo Scientific Potassium Dichromate Solution Standards are NIST-traceable for qualifying instrument performance. Qualify your spectrophotometer's performance with NIST-traceable Thermo Scientific Potassium Dichromate Solution Standards for photometric accuracy.
h: Holmium oxide solution verification standard for UV-Vis spectrophotometers - Thermo Scientific NIST-traceable Holmium Oxide Solution Standard verifies wavelength accuracy of UV-Visible spectrophotometers.
i: Evolution Fiber Optic Couplers for UV-Vis spectrophotometers - Maintain the integrity of your samples and add remote sampling capabilities to your laboratory with support for fiber optic probes. Thermo Scientific Evolution Fiber Optic Couplers, for use with the Evolution Spectrophotometers, allow you to measure your samples with one of our fiber optic probes or with any third-party probe equipped with standard SMA connectors. The probe brings the light to any sample container, so you don't waste time bringing the sample to the instrument in a cuvette or test tube.
j: Holmium and didymium glass standards for UV-Vis spectrophotometers - Thermo Scientific Wavelength Calibration Filter Sets consist of Holmium and Didymium solid filters traceable to international standards.
k: SPECTRONIC Thermal Printer for SPECTRONIC 200 Spectrophotometer - External Thermal Printer for SPECTRONIC 200/200E
l: SPECTRONIC Spectrophotometer Standards - Check the performance and accuracy of your spectrophotometer right in your own laboratory using the Thermo Scientific SPECTRONIC Spectrophotometer Standards. This kit is as easy to use as a standard cuvette and contains:
m: Toluene in Hexane Solution Standard for UV-Vis spectrophotometers - Thermo Scientific traceable Toluene in Hexane Solution Standard ensures accurate wavelength and absorbance results by verifying spectrophotometer resolution.
n: Evolution Peltier Accessories for UV-Vis spectrophotometers - Temperature control and sample stirring are important for many UV-Visible analysis methods. Peltier thermostatted accessories outperform traditional heater/chiller units to deliver superior performance with compact integrated design, simple set-up, and fast, precise temperature control. Thermo Scientific Peltier Accessories for the Evolution 200 Series and Evolution 350 UV-Vis Spectrophotometers provide thermostatting and ramping with precise temperature control between 0 °C and 110 °C.
For Research Use Only. Not for use in diagnostic procedures.