Search
Lipofectamine® LTX Reagent is a proprietary, animal-origin free formulation for the transfection of DNA into eukaryotic cells with low cytotoxicity. This reference provides a recommended procedure to transfect plasmid DNA into K562 Human Myelogenous Leukemia Cells (ATCC Cat. No. CCL243) using Lipofectamine™ LTX Reagent (Cat. No. 15338-100).
Important Guidelines for Transfection
Follow these important guidelines when transfecting DNA into K562 cells using Lipofectamine® LTX Reagent:
Materials Needed
Have the following reagents on hand before beginning:
To transfect K562 cells in different tissue culture formats, vary the amounts of Lipofectamine® LTX Reagent, DNA, cells, medium and PLUS™ Reagent used in proportion to the relative surface area, as shown in the table (amounts given on a per well basis).
| Culture vessel | Surface area per well1 | Volume plating medium | Cells per well | Volume dilution medium2 | DNA | Lipofectamine® LTX Reagent | PLUS™ Reagent |
|---|---|---|---|---|---|---|---|
| 96-well | 0.3 cm2 | 100 μl | 2.5 x 104 | 20 μl | 100 ng | 0.25 - 0.55 μl | 0.1 μl |
| 48-well | 1 cm2 | 200 μl | 5 x 104 | 40 μl | 200 ng | 0.5 - 1.1 μl | 0.2 μl |
| 24-well | 2 cm2 | 500 μl | 1 x 105 | 100 μl | 500 ng | 1.25 - 2.75 μl | 0.5 μl |
| 12-well | 4 cm2 | 1 ml | 2 x 105 | 200 μl | 1 μg | 2.5 - 5.5 μl | 1.0 μl |
| 6-well | 10 cm2 | 2 ml | 5 x 105 | 500 μl | 2.5 μg | 6.25 - 13.75 μl | 2.5 μl |
1 Surface areas may vary depending on the manufacturer.
2 If the volume of Lipofectamine® LTX Reagent is too small to dispense accurately, and you cannot pool dilutions, predilute Lipofectamine® LTX Reagent 10-fold in Opti-MEM® I Reduced Serum Medium, and dispense a 10-fold higher amount (should be at least 1.0 μl per well). Discard any unused diluted Lipofectamine® LTX Reagent.