Search
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
First check the percentage of your agarose gel. A higher percentage will help you to resolve smaller molecular weights while a lower percentage will help you to resolve larger molecular weights.
Check the power supply to ensure current is being passed through the gel. The running buffer composition must match the gel chemistry.
Usually, fuzzy bands are caused by incorrect buffer solutions. Check the pH and salt concentrations in the running and gel buffers. Fuzzy bands can also be caused by excess salt in the samples or by old gels.
We would recommend running the gel for longer or choosing a different gel that will have higher resolution in the size range you are targeting.
Do not run the gels longer than 60 minutes for the single comb gels or 25 minutes for the double comb gels. Ions are depleted and longer run times will damage the gel. Do not run E-Gel® 48 Agarose Gels longer than 30 minutes or E-Gel® 96 Agarose Gels longer than 20 minutes.
The conformation of plasmid DNA will affect its mobility on a gel. Plasmid DNA can be supercoiled (native 3D conformation), nicked circular (nick in one DNA strand) or linear (nicks in both DNA strands). You may see all three forms on an agarose gel. Supercoiled DNA runs the fastest and will show up lowest on the gel, linear DNA runs in the middle of the gel, and nicked circular DNA shows up at the top of the gel, since it migrates the slowest.
While we recommend storage at room temperature, these gels will still be usable. Bring the gels to room temperature prior to the run for optimal conditions.
Potential issues are:
Please ensure that you have not overloaded the well and that the wells were not damaged during comb removal.
Check the E-Gel® cassette and copper contacts. You can try replacing the gel cassette with a fresh gel cassette to see whether this fixes the problem. A cold cassette or improper operating conditions can also lead to this failure. Cassettes should be at room temperature for use; avoid storing at 4°C.
Here are some suggestions:
Please check the cord you are using for the iBase™ Power System. Oftentimes, the power cords for the PowerBase™ v.4 system and the E-Gel® Safe Imager™ Real-Time Transilluminator get mixed up. If this is the case, the blue light will not come on even though the fan and LED light will operate. You will need to use the iBase™ cord, which should say 48 V on it.
Here are some common reasons why your gel is not running properly:
Here are some possibilities and suggestions to resolve the problem:
We do not currently offer software that is compatible with Mac® computers. Please install the software on a 32-bit or 64-bit PC running either a Windows® XP Professional or Windows® 7 Professional operating system.
This error can occur if there is other software running in the background, like an Internet Explorer® browser, or if there is an antivirus program running that is limiting his ability to access some files.
To fix this error, try to:
Depress the activation button for at least 2 seconds, and the imager will transilluminate for 5 minutes rather than 30 seconds.
1) Check that the camera hood or activator key is in place to make sure the base is not deactivated.
2) Light bases will "time out," but the power indicator light may stay illuminated. Turn the base off, then back on.
Follow recommended DNA dilutions and leave the gel to cool down on the bench or for a few minutes in the fridge. Please check troubleshooting tips provided in the manual.
If a slight turbidity develops, the fine precipitate can be dissolved by autoclaving for 5 minutes at 110°C. Do not autoclave in the container supplied. This treatment has no deleterious effect on the buffering properties of TBE.
Here are some suggestions for your experiments:
You can use the Reverse E-Gel® Program to run the band back into the collection gel.
Please see below the top ten ways to increase sensitivity of your Northern hybridizations:
1) Increase the amount of RNA loaded in each lane (up to 30 mg). 2) Use poly(A) RNA instead of total RNA; 10 mg of poly(A) RNA is ~300–350 mg total RNA (3–5%). 3) Switch to ULTRAhyb® Ultrasensitive Hybridization Buffer. 4) Switch from DNA to RNA probes. 5) Use downward alkaline capillary transfer. 6) Use an optimal hybridization temperature. 7) Use a freshly synthesized probe. 8) Use a high specific activity probe (10^8 to 10^9 cpm/mg). 9) Increase exposure time (it can take up to 3 days to see low-abundance messages with radiolabeled probes). 10) Follow the manufacturer’s recommendations to crosslink the RNA to the membrane.
Read more about these suggestions here.
For RNA probes on DNA or RNA targets: Autoclave the membrane in a bottle containing 0.1% SDS solution for 15 minutes. Repeat if necessary. For DNA probes on DNA targets only: You can use the same protocol used for RNA probe stripping. Another option is alkaline denaturation. Incubate the membrane with 400 mM NaOH for 30 minutes, then wash with 0.1% SDS for 15 minutes. These stripping methods should work for 2–3 stripping procedures. However, nucleic acids will gradually be removed from the blot.
rRNA makes up ~80% of total RNA samples. When 10 µg of total RNA is loaded into a Northern gel lane, the 18S and 28S rRNA bands contain 2–6 µg RNA each. This amount of nucleic acid can nonspecifically trap probe as well as bind complementary sequence. Probe trapping by rRNA can be reduced by using the minimal amount of probe, and by labeling only sequence complementary to mRNA. Transfer using a basic buffer can prevent trapping. Finally, you can use a high hybridization and wash temperature to minimize cross hybridization to rRNA.
Residual RNA could be due to:
Poor signal could be a result of the following:
There are several types of background, and each can have a different cause:
1) Blotchy signal across the membrane: This can be caused by a membrane of poor quality, one that has dried out, or one that has been mishandled (e.g., oil from human skin, powder from gloves). Use high quality nylon membrane that has not previously been handled and use forceps to handle the membrane from the edges. Blotchiness can also be caused by uneven distribution of the hybridization reagents. Do not pipette probe directly onto the membrane in hybridization solution; dilute it into the hybridization solution first.
2) A smear through the lane: Hybridization conditions that are substantially below the optimum for a given probe can lead to high lane-specific background and/or substantial cross-hybridization. Start with a high hybridization temperature and slowly decrease the temperature until a specific signal is obtained. High probe concentrations, especially for nonisotopic probes, can also cause lane-specific background. Use 10 pM nonisotopically labeled DNA probes and 0.1 nM nonisotopically labeled RNA probes.
3) Speckling across the membrane: Probe preparations with poor incorporation (or where unincorporated nucleotides have not been removed) can cause speckling on the membrane. Check probe quality and remove unincorporated nucleotides. Particulates in probe preparations or hybridization buffer (e.g., when not completely in solution) can also cause speckling on the membrane. Ensure that these reagents are in solution, and consider centrifuging in a microfuge or low-speed centrifuge, or filtering the solutions through a 0.22 µm filter to remove particulates.
If you see high background that is not associated with the lanes, this could be due to:
The following reasons could have led to cross-hybridization:
Incomplete transfer is often caused by short-circuiting. Strips of Parafilm® sealing film around the outside edges of the gel can prevent this.
Large RNA species may not transfer well because of their size. A basic transfer buffer (e.g., NorthernMax® One-Hour Transfer Buffer) will partially shear the RNA so that larger RNA species transfer more efficiently.
Check RNA transfer by including ethidium bromide in RNA samples or staining the gel in ethidium bromide after transfer and viewing your gel under UV light. RNA markers are invaluable to demonstrate whether large RNAs have fully transferred. Our Ambion® Millennium™ Markers are especially useful for this purpose, since they include transcripts at 1,000 nt intervals from 0.5 to 9 kb.
For Research Use Only. Not for use in diagnostic procedures.