On this page:
XPS depth profiling for thin films, coatings, and multi-layer structures
The Thermo Scientific Hypulse Surface Analysis System supplements traditional ion-beam methods with femtosecond laser ablation (fs-LA) technology to offer outstanding X-ray photoelectron spectroscopy (XPS) depth profiling capabilities for thin films, coatings, and multi-layer structures. Fs-LA XPS depth profiling provides even more accurate, efficient, and versatile analysis from the surface to interfaces deep with the sample. This helps you optimize fabrication and the properties of technologically important materials and devices, develop new multi-functional materials, and gain a deeper understanding of material failures.
Unlock new depths of material analysis with femtosecond laser ablation XPS
Fs-LA XPS delivers accurate, reliable surface and interface analysis without the risk of chemical damage to the sample. With this cutting-edge technique, you can quickly, efficiently, and precisely measure technologically important materials and devices to better understand their structure.
Designed for precision and accuracy, the Hypulse System allows you to explore the intricate layers of diverse materials while preserving their chemistry during the analysis. Whether you are in the electronics, metallurgy, or surface coatings industries, it provides fast, detailed, and accurate analysis.
Get the eBook
Learn more about the Hypulse System and review case studies.
Get the eBook
Revolutionizing XPS depth profiling for materials science
Accurate and efficient chemical analysis is essential for materials science research labs, academic institutions, and industries such as semiconductor manufacturing, polymer engineering, coating development, and the development of new multifunctional materials. Traditional ion gun-based methods for X-ray photoelectron spectroscopy depth profiling can have issues with certain materials, which results in data that is not completely representative of the sample.
Webinar: Explore damage-free XPS depth profiling with the Hypulse System
Watch on demand to learn how the Hypulse System combines femtosecond laser ablation with ion beam milling to help you analyze materials without chemical damage.
Hypulse Surface Analysis System benefits
Achieve more accurate analysis
By combining monatomic ion beam etching, cluster ion beam etching, and fs-LA depth profiling, the Hypulse System can provide accurate results for more materials than was previously possible.
Enhance productivity
Fs-LA depth profiling can perform multiple profiles on a material in the time that one sample can be processed using ion beam profiling.
Gain a more comprehensive understanding of materials
The Hypulse System can help you practically measure depth profiles that extend beyond 10 µm into the sample. Buried layers that would take significant time to discover using ion beam profiling can be accessed more easily through fs-laser ablation depth profiling.
Optimize your research for different types of materials
Rather than choosing between cluster ions or monatomic ions, the Hypulse System’s laser settings can be tuned to the material, making it easier to profile complex composite samples.
Hypulse Surface Analysis System features
Integrated fs-laser ablation system
The Hypulse System is built around a fully integrated 1,030 nm fs-laser system and high-quality optical components. The laser is focused on the analysis position and can be attenuated through Thermo Scientific Avantage Software. The instrument is a class 1 laser system, so no special laser requirements are necessary for the laboratory.
MAGCIS ion source
To complement the fs-LA system, the Thermo Scientific MAGCIS Dual Beam Ion Source for XPS Instruments is included as standard. Automatic source optimization and gas handling ensure excellent performance and experimental reproducibility.
High-performance surface analysis
The Hypulse System is a high-performing XPS  instrument and includes additional analytical techniques such as REELS and ISS to help you get the most information from your samples.
XPS insulator analysis
The patented dual-beam flood source couples low-energy ion beams with very low-energy electrons (less than 1 eV) to prevent sample charging during analysis, which eliminates the need for charge referencing and makes analyzing data from insulating samples easy and reliable.
Optional XPS sample holders
Expand your possibilities with available specialist sample holders for angle-resolved XPS, sample bias measurements, sample heating, or inert transfer from a glove box.
Avantage Software
Instrument control, data processing, and reporting are all controlled by Avantage Software. The software includes Knowledge View, a comprehensive source of tutorials and reference material that guides you through analysis.
Correlative XPS imaging and surface analysis workflow
Correlate XPS data with SEM images through the correlative imaging and surface analysis (CISA) workflow using Thermo Scientific Maps Software.
Hypulse Surface Analysis System specifications
Analyzer type
  • 180º, double-focusing, hemispherical analyzer with 128-channel detector
X-ray source
  • Monochromated, micro-focused, low-power Al K-alpha X-ray source
X-ray spot size
  • 10 to 400 µm (adjustable in 5 µm steps)
Laser ablation system
  • Fully integrated 1,030 nm pulsed fs-laser source
  • Tunable pulse energy up to 1 mJ
  • Computer-controlled attenuation to ensure optimum ablation conditions
  • Beam imaging module for system calibration and alignment
Ion depth profiling
  • MAGCIS dual-mode ion source
Additional included analytical methods
  • Bi-polar analyzer power supply and helium gas input for ion source for ISS (ion scattering spectroscopy)
  • High-voltage upgrade to electron flood source for REELS (reflection electron energy loss spectroscopy)
Maximum sample area
  • 60 x 60 mm
Maximum sample thickness
  • 20 mm
Vacuum system
  • Two turbo molecular pumps with automated titanium sublimation pump and backing pump
Optional accessories
  • UV photoelectron spectroscopy (UPS)
  • Sample tilt module
  • NX sample heating module
  • Sample bias module
  • Vacuum transfer module
XPS analysis resources
Hypulse Surface Analysis System documentation
Calibrating ablation rates for femtosecond-laser XPS depth profiling with the Hypulse Surface Analysis System
Hypulse Surface Analysis System
The evolution of XPS depth profiling
XPS depth profiling of coated nitrided steel using femtosecond laser ablation
XPS depth profiling of advanced solar cells with femtosecond laser ablation
Polymer analysis using femtosecond-laser-ablation depth profiling
Exploring the surface in depth with XPS analysis
Hypulse Surface Analysis System videos
For Research Use Only. Not for use in diagnostic procedures.