Search
By using the correct reagent, you can increase or decrease retention and control selectivity, resolve complex ionic mixtures without using ion exchange columns, and improve peak symmetry. Also see our ultrapure solvents for high-sensitivity HPLC and LC-MS analysis, and special reagents for amino acid and peptide applications.
We offer a variety of HPLC detection reagents that enable visualization of your compound of interest. These reagents can improve both selectivity and specificity, and can be used before or after separation.
Quickly and efficiently analyze highly charged acidic or basic compounds using reversed phase techniques. The reagents form stable complexes with these compounds, enabling resolution by reversed phase HPLC.
HPLC and spectrophotometric grade solvents are ultrapure for maximum LC/MS sensitivity. They can extend column lifetime, eliminate variability and reduce handling.
We offer sequencing-grade 6N HCl and amino acid standard H for protein hydrolysates as well as ninhydrin for amino acid detection.
Various retention standards and calibration mixtures that provide reference elution profiles in reverse-phase HPLC.
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.