Search
The Thermo Scientific iCAP MSX ICP-MS instrument incorporates easy-to-use technology that delivers right-first-time analysis. It enables you to achieve greater workflow efficiency without the compromise of maintenance downtime. From sample input to analytical output, it fulfills the most demanding requirements with effortless efficiency.
Simple sample introduction
With an optimized sample introduction system, including a built-in solution for diluting high matrix containing samples using argon gas dilution (AGD), the system helps to ensure consistent and stable analytical performance no matter the sample type.
Optimized for matrix
The Intelligent Matrix Handling feature reduces the unnecessary introduction of matrix during sample uptake and wash out, increasing the time between instances of interface maintenance.
Highly sensitive
The IntelliLens uses advanced tuning, which applies optimized voltage settings for every individual analyte to provide exceptional sensitivity across the entire mass range.
Effective interference removal
Comprehensive interference removal is accomplished using the QCell collision/reaction cell (CRC), enabling effective suppression of polyatomic interferences based on a two-stage interference removal mechanism featuring an automatically adjusted low mass cut-off.
For Research Use Only. Not for use in diagnostic procedures.