Oncomine Reporter: A Next-Generation Sequencing Bioinformatics Solution for Oncology Research
Next-generation sequencing (NGS) reporting & analysis challenges in oncology research
Generating clear and concise oncology research reports that contain accurate information in a timely manner can be challenging. As the number of biomarker-driven therapy approvals increases, managing the targeted therapies for each type of cancer and updating this information from multiple sources becomes difficult and impractical.
Today’s challenges include:
  • A growing number of oncology biomarkers with potential utility
  • Multiple global sources of information that are not standardized
  • Variants identified via next-generation sequencing need to be accurately associated with relevant information
Watch a quick overview of Oncomine Reporter
Oncomine Reporter simplifies NGS reporting for cancer research
Oncomine Reporter is a reporting software solution developed with a streamlined three-step workflow. It delivers easy access to vital information which enables the contextual investigation of sample-specific variants with respect to labels, guidelines, current clinical trials, and peer reviewed literature obtained from public sources.
On Demand Webinar: Oncomine Reporter - Integrated Reporting for Solid Tumor and Myeloid Somatic Variants
Advancements in NGS technology have resulted in streamlined, near end-to-end processes requiring minimal touch points by the laboratory technicians. Two remaining challenges for many researchers are the handling and analysis of data generated by NGS and generating intuitive reports to interpret the results. In this discussion, we will cover a clinical laboratory's historic usage of reporting software, including how they have advanced over the years.
Dr. David Seidman, Ph.D., MB (ASCP)cm
Scientific Director, Molecular Diagnostics and Serology Laboratories
Sentara Healthcare Laboratory Services
Features of Oncomine Reporter
User-friendly software
Easily create reports to your specifications, on your timeline, in just three steps
Built for NGS
NGS support for solid tumor hematological cancers including SNVs, indels, fusions, CNVs with TMB, LOH,
MSI, and HRR
Customizable reports
Get all relevant information in a concise and customized report
Global content
Curated global information from multiple public sources, delivered specifically for your use
From variants to a final NGS report in three steps
Step 1: Review
Get results from solid tumor, liquid biopsy, hematological cancer research or cytogenetics runs
Step 2: Filter
Customize results by selecting specific samples as well as filtering on data sources, location, and other parameters
Step 3: Report
Design templates to show the data that is most valuable to you in templates that are available in 10 languages
Flexible report customization options
Oncomine Reporter has an easy-to-use interface with filtering options based on data source, cancer type, and clinical trial location. Role-based user profiles provide the flexibility to standardize workflows for operators in laboratory environments.
The report builder enables you to select and order the sections of the report. This flexibility lets you create customized reports that contain only summary information or as much detail as you need.
Ordering information
Catalog # Name Size Price Qty
A34298 Each
91.25
Ready to speak with a Thermo Fisher representative?
Want to learn more?  We are happy to answer any questions you have about Oncomine Reporter.