RiboMinus™ Kits for RNA-Seq Banner
RiboMinus Kits for RNA-Seq
RiboMinus™ Kits for RNA-Seq
Enrichment of the true whole transcriptome through selective depletion of ribosomal RNA.
How RiboMinus Works
photo of RiboMinus rRNA depletion kit box, vials of magnetic beads, and reagent bottles
RiboMinus is a novel technology that utilizes Locked Nucleic Acid to selectively deplete up to 99.9% of ribosomal RNA.
  • Learn how RiboMinus Technology works
RiboMinus Improves Sequencing Data
RiboMinus™ Improves Sequencing Data
The Ribominus Eukaryote and Plant Kits for RNA-Seq provide a method for true whole transcriptome isolation through a unique method employing selective depletion of ribosomal RNA.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments