Search
TaqMan® Protein Assays are designed for highly sensitive and specific analysis of protein expression in small samples. Predesigned assays have been developed for the pluripotency markers OCT4, NANOG, SOX2, and LIN28 in human stem cells as well as the ubiquitous human proteins CSTB and ICAM1.
Stem Cell Marker Proteins Readily Detected In 100–500 Cells Using TaqMan® Protein Assays.
Related Documents