Search
Elevate your respiratory research endeavors with Applied Biosystems high-sensitivity real-time PCR products. Our solutions enable the comprehensive analysis of various respiratory pathogens, encompassing viruses, bacteria, and fungi, all with exceptional analytical sensitivity. By harnessing the power of our molecular methods, you can overcome the inherent challenges associated with immunoassay-based and culture-based techniques. Experience efficiency, informative results, and reliability as you delve into the intricacies of respiratory pathogen studies.
|
Single-tube assay |
Plates |
Cards |
OpenArray plates |
Formats |
Individual tubes or assay tube sets in a variety of reaction sizes |
96-and 384-well plates with assays pre-spotted into the wells |
384-well microfluidic cards with assays pre-spotted |
3,000 through-holes with assays pre-spotted |
Assay type |
Singleplex or Spectral Multiplex |
Spectral Multiplex |
Singleplex |
Spatial-Multiplex |
Typical # of targets |
1 to 5 |
4 to 24 |
24 or 48 |
18 or 56 |
Use case |
For smaller panels in any volume (1 to 1,000s) |
For mid-size panels in mid-throughput (10–50) |
For large panels in low throughput (10s) |
For mid to large panels in high throughput (48–1,000s) |
Your advantage |
High flexibility for low throughput |
Supports automation for medium throughput |
Convenience—no robotics needed |
Higher throughput for routine applications or large projects |