Lab Automation Incubators
Automated incubators and storage solutions
Laboratory equipment manufacturers and researchers worldwide depend on highly reliable Thermo Scientific Cytomat automated incubators and microplate storage systems. With more than 50 years of experience in CO2 incubator technology, the robot-accessible Cytomat series is the established leader in the field.
Featured automated incubator and storage categories
The right choice for your application at 37°C up to 50°C (70°C) in many capacities.
Ideal environment for storage and incubation of your assays in a temperature range from 4°C up to 50°C (70°C) with multiple capacities.
Automated tower shaker solutions
The Thermo Scientific Cytomat Tower Shaker is the only solution that provides true orbital shaking. It is ideal for applications requiring sample agitation and cells that need to be kept in suspension.
  • Easy 3-step setup of Tower Shaker – install/secure by lock screw to the ceiling/plug-in for communication
  • Active microplate clamp mechanism secures plate and lid separately; zero microplate abrasions during shaking
  • Mix and match standard and Tower Shaker stackers for maximum flexibility
  • Inductive wear-free driving mechanism reduces particle emissions and belt changes
  • Superior shaking technology with individual speed settings for each Tower Shaker from 300-1200 rpm with oscillating amplitude of 2 mm or 3 mm offers a wide range of application setups
The Thermo Scientific Cytomat 2 C4xx-LiN ToS automated incubator with a capacity up to 32 microplates is the ideal solution for applications with a temperature range of 4-70°C.
Featured videos
Thermo Scientific Cytomat 10 Series
This video shows the Cytomat 10 automated incubator series combines superior environmental control, fast access times and versatility, making it the ideal choice for cell-based workflows requiring larger capacity incubation.
Video Player is loading.
Current Time 0:00
Duration 2:57
Loaded: 0%
Stream Type LIVE
Remaining Time 2:57
1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
    • en (Main), selected
      1. 現在再生中
        次の動画Thermo Scientific Cytomat 10 Series
        This video shows the Cytomat 10 automated incubator series combines superior environmental control, fast access times and versatility, making it the ideal choice for cell-based workflows requiring larger capacity incubation.
      2. 現在再生中
        次の動画Thermo Scientific™ Cytomat™ 2 C-LiN Series Automated Incubator and Storage Devices
        The next generation of advanced incubation and storage solutions for cell culture applications.
      3. 現在再生中
        次の動画Thermo Scientific Cytomat 2 C4xx-LiN ToS-Tower-Shaker product video
        Automated incubator with true orbital shaking capability based on individual Tower Shaker technology with different shaking amplitudes and frequencies.