Catalysis research
Involved in the processing of over 80% of all manufactured products, catalysts are a critical aspect of modern industry. Heterogeneous nanoparticle catalysts, in particular, are important for a number of modern, environmentally friendly processes such as the production of hydrogen fuel and are found ubiquitously in automotive catalytic converters. As catalysts accelerate production rates and lower temperature requirements for relevant reactions, they significantly reduce the energy needed to perform a given process and/or produce a product of interest.
Nanoparticle catalysis
The morphology, distribution, size, and chemical composition of nanoparticles are crucial for their catalytic efficiency. Scanning transmission electron microscopy (S/TEM) combined with energy-dispersive X-ray spectroscopy (EDS) has proven to be a valuable research tool for the direct observation and quantification of this information. Additionally, high-performance scanning electron microscopy (SEM) tools take excellent images of beam-sensitive catalyst materials under low-beam-energy and low-beam-current conditions without causing sample damage.
Thermo Fisher Scientific provides a range of instrumentation ideally suited for the characterization of catalyst nanoparticles. We also offer a suite of software tools that allow you to automate your workflow, generating high-resolution, large-area nanoparticle data for a holistic overview of your catalyst.
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
Platinum nanoparticles and cobalt nanoparticles, used to improve catalytic efficiency, imaged with high resolution EDS
High-resolution EDS maps of a beam-sensitive material used for photocatalytic processes (C3N4(Co)-Pt). The catalyst uses the synergistic behavior of platinum and cobalt nanoparticles to improve catalytic efficiency. Data courtesy of Prof. ShengChun Yang, Xi’an Jiaotong University, China.
Resources
On-Demand Webinar: Spectra 200 S/TEM: A workhorse for catalyst characterisation
Watch this webinar to learn advanced catalyst characterisation methods using TEM, how tools like the Spectra 200 S/TEM are used in the catalyst industry and why Haldor Topsøe chooses the Spectra 200 S/TEM.
Watch webinar
On-Demand Webinar: Quantitative structural analysis using STEM HAADF-iDPC
This webinar will give an introduction to the relevance of materials in catalysis, the challenges in their synthesis and their characterisation at atomic level.
Watch webinar
High-resolution TEM imaging of Pt-Rh catalyst nanoparticles enabled by the Thermo Scientific Ceta 16M Camera, revealing the crystalline lattice structure of nanoparticles. Sample courtesy: Prof. B. Gorman and Prof. R. Richards, Colorado School of Mines.
On-Demand Webinar: Spectra 200 S/TEM: A workhorse for catalyst characterisation
Watch this webinar to learn advanced catalyst characterisation methods using TEM, how tools like the Spectra 200 S/TEM are used in the catalyst industry and why Haldor Topsøe chooses the Spectra 200 S/TEM.
Watch webinar
On-Demand Webinar: Quantitative structural analysis using STEM HAADF-iDPC
This webinar will give an introduction to the relevance of materials in catalysis, the challenges in their synthesis and their characterisation at atomic level.
Watch webinar
High-resolution TEM imaging of Pt-Rh catalyst nanoparticles enabled by the Thermo Scientific Ceta 16M Camera, revealing the crystalline lattice structure of nanoparticles. Sample courtesy: Prof. B. Gorman and Prof. R. Richards, Colorado School of Mines.
Applications
Process Control_Thumb_274x180_144DPI
Control de proceso
La industria moderna exige un alto rendimiento con una calidad superior, un equilibrio que se mantiene a través de un control de procesos sólido. Las herramientas SEM y TEM con software de automatización exclusivo proporcionan información rápida y multiescala para la supervisión y la mejora de procesos.
Fundamental Materials Research_R&D_Thumb_274x180_144DPI
Investigación sobre materiales fundamentales
Se investigan nuevos materiales a escalas cada vez más pequeñas para lograr el máximo control de sus propiedades físicas y químicas. La microscopía electrónica proporciona a los investigadores información clave sobre una amplia variedad de características materiales a escala nanométrica.
Style Sheet for Komodo Tabs
Techniques
Preparación de muestras (S)TEM
Los microscopios DualBeam permiten la preparación de muestras ultrafinas de alta calidad para el análisis (S)TEM. Gracias a la automatización avanzada, los usuarios con cualquier nivel de experiencia pueden obtener resultados de nivel experto para una amplia gama de materiales.
Más información ›
Caracterización de materiales en 3D
El desarrollo de materiales suele requerir caracterización en 3D en varias escalas. Los instrumentos DualBeam permiten el corte en secciones en serie de grandes volúmenes y la posterior adquisición de imágenes SEM a escala de nanómetro, las cuales se pueden procesar en reconstrucciones 3D de la muestra de alta calidad.
Más información ›
Análisis elemental EDS
EDS proporciona información de composición vital sobre las observaciones de microscopio electrónico. En concreto, nuestros exclusivos sistemas de detectores Super-X y Dual-X añaden opciones para mejorar el rendimiento y/o la sensibilidad, permitiendo optimizar la adquisición de datos para cumplir con sus prioridades de investigación.
Más información ›
Tomografía EDS en 3D
La investigación de materiales modernos depende cada vez más del análisis a nanoescala en tres dimensiones. La caracterización en 3D, incluidos los datos de composición para el contexto químico y estructural completo, es posible con EM en 3D y espectroscopia de rayos X dispersiva.
Más información ›
Espectroscopia de energía dispersiva
La espectroscopía de energía dispersiva (EDS) recopila información elemental detallada junto con imágenes de microscopía electrónica, proporcionando un contexto de composición esencial para las observaciones de EM. Con EDS, se puede determinar la composición química desde barridos de superficie rápidos y holísticos hasta átomos individuales.
Más información ›
Asignación elemental a escala atómica con EDS
El EDS de resolución atómica proporciona un contexto químico incomparable para el análisis de materiales al diferenciar la identidad elemental de los átomos individuales. Cuando se combina con TEM de alta resolución, es posible observar la organización precisa de los átomos en una muestra.
Más información ›
ColorSEM
Mediante la utilización de EDS en tiempo real (espectroscopia de rayos X por dispersión de energía) con cuantificación en tiempo real, la tecnología ColorSEM transforma las imágenes SEM en una técnica de color. Cualquier usuario puede adquirir datos elementales de forma continua para obtener información más completa que nunca.
Más información ›
Adquisición de imágenes con HRSTEM y HRTEM
La microscopía electrónica de transmisión es muy valiosa para caracterizar la estructura de nanopartículas y nanomateriales. EL TEM y STEM de alta resolución permiten obtener datos de resolución atómica junto con información sobre la composición química.
Más información ›
Adquisición de imágenes de contraste de fase diferencial
La investigación electrónica moderna se basa en el análisis a nanoescala de las propiedades eléctricas y magnéticas. El contraste de fase diferencial STEM (DPC-STEM) puede obtener imágenes de la fuerza y la distribución de los campos magnéticos en una muestra y mostrar la estructura de dominio magnético.
Más información ›
Adquisición de imágenes de muestras calientes
El estudio de los materiales en condiciones reales suele implicar el trabajo a altas temperaturas. El comportamiento de los materiales cuando se recristalizan, derriten, deforman o reaccionan ante el calor se puede estudiar in situ con la microscopía electrónica de barrido o con las herramientas DualBeam.
Más información ›
SEM ambiental (ESEM)
El SEM ambiental permite que se adquieran imágenes de los materiales en su estado nativo. Esto es ideal para investigadores académicos e industriales que necesitan probar y analizar muestras húmedas, sucias, reactivas, con liberación de gases o que no son compatibles con el vacío.
Más información ›
Espectroscopía de pérdida de energía de electrones
La investigación en ciencias de los materiales se beneficia de la EELS de alta resolución para una amplia gama de aplicaciones analíticas. Esto incluye asignación elemental de alto rendimiento, alta relación señal-ruido, así como sondeo de estados de oxidación y fonones de superficie.
Más información ›
Corte transversal
El corte transversal proporciona una visión adicional, ya que descubre información de la subsuperficie. Los instrumentos DualBeam tienen columnas FIB para poder realizar el corte transversal con alta calidad. Con la automatización, se puede realizar el procesamiento de muestras de alto rendimiento sin supervisión.
Más información ›
Experimentación in situ
La observación directa y en tiempo real de los cambios microestructurales con microscopía electrónica es necesaria para comprender los principios subyacentes de los procesos dinámicos como la recristalización, el crecimiento del grano y la transformación de fases durante el calentamiento, refrigeración y humectación.
Más información ›
Análisis de partículas
El análisis de partículas juega un papel vital en la investigación de nanomateriales y el control de calidad. La resolución a escala nanométrica y la adquisición de imágenes superiores de microscopía electrónica se pueden combinar con software especializado para la rápida caracterización de polvos y partículas.
Más información ›
SIMS
El detector TOF-SIMS (espectrometría de masas de iones secundaria de tecnología de tiempo de vuelo) para microscopía electrónica de tecnología barrido de haz de iones (FIB-SEM) permite la caracterización analítica de alta resolución de todos los elementos de la tabla periódica, incluso a bajas concentraciones.
Más información ›
Análisis de escala múltiple
Los novedosos materiales se deben analizar a una resolución cada vez mayor, manteniendo el contexto más amplio de la muestra. El análisis de escala múltiple permite la correlación de varias herramientas y modalidades de obtención de imágenes, tales como microTC de rayos X, DualBeam, PFIB láser, SEM y TEM.
Más información ›
Espectroscopía de fotoelectrones de rayos X
La espectroscopía de fotoelectrones de rayos X (XPS) permite el análisis de superficie, proporcionando la composición elemental, así como el estado químico y electrónico de los 10 nm principales de un material. Con la realización de perfiles de profundidad, el análisis XPS se extiende a la visión de composición de las capas.
Más información ›
Flujo de trabajo de partículas automatizado
El flujo de trabajo de nanopartículas automatizado (APW) es un flujo de trabajo de microscopio electrónico de transmisión para el análisis de nanopartículas que proporciona adquisición de imágenes de área extensa y de alta resolución, además de adquisición de datos en nanoescala, todo ello con un procesamiento sobre la marcha.
Más información ›
Preparación de muestras (S)TEM
Los microscopios DualBeam permiten la preparación de muestras ultrafinas de alta calidad para el análisis (S)TEM. Gracias a la automatización avanzada, los usuarios con cualquier nivel de experiencia pueden obtener resultados de nivel experto para una amplia gama de materiales.
Más información ›
Caracterización de materiales en 3D
El desarrollo de materiales suele requerir caracterización en 3D en varias escalas. Los instrumentos DualBeam permiten el corte en secciones en serie de grandes volúmenes y la posterior adquisición de imágenes SEM a escala de nanómetro, las cuales se pueden procesar en reconstrucciones 3D de la muestra de alta calidad.
Más información ›
Análisis elemental EDS
EDS proporciona información de composición vital sobre las observaciones de microscopio electrónico. En concreto, nuestros exclusivos sistemas de detectores Super-X y Dual-X añaden opciones para mejorar el rendimiento y/o la sensibilidad, permitiendo optimizar la adquisición de datos para cumplir con sus prioridades de investigación.
Más información ›
Tomografía EDS en 3D
La investigación de materiales modernos depende cada vez más del análisis a nanoescala en tres dimensiones. La caracterización en 3D, incluidos los datos de composición para el contexto químico y estructural completo, es posible con EM en 3D y espectroscopia de rayos X dispersiva.
Más información ›
Espectroscopia de energía dispersiva
La espectroscopía de energía dispersiva (EDS) recopila información elemental detallada junto con imágenes de microscopía electrónica, proporcionando un contexto de composición esencial para las observaciones de EM. Con EDS, se puede determinar la composición química desde barridos de superficie rápidos y holísticos hasta átomos individuales.
Más información ›
Asignación elemental a escala atómica con EDS
El EDS de resolución atómica proporciona un contexto químico incomparable para el análisis de materiales al diferenciar la identidad elemental de los átomos individuales. Cuando se combina con TEM de alta resolución, es posible observar la organización precisa de los átomos en una muestra.
Más información ›
ColorSEM
Mediante la utilización de EDS en tiempo real (espectroscopia de rayos X por dispersión de energía) con cuantificación en tiempo real, la tecnología ColorSEM transforma las imágenes SEM en una técnica de color. Cualquier usuario puede adquirir datos elementales de forma continua para obtener información más completa que nunca.
Más información ›
Adquisición de imágenes con HRSTEM y HRTEM
La microscopía electrónica de transmisión es muy valiosa para caracterizar la estructura de nanopartículas y nanomateriales. EL TEM y STEM de alta resolución permiten obtener datos de resolución atómica junto con información sobre la composición química.
Más información ›
Adquisición de imágenes de contraste de fase diferencial
La investigación electrónica moderna se basa en el análisis a nanoescala de las propiedades eléctricas y magnéticas. El contraste de fase diferencial STEM (DPC-STEM) puede obtener imágenes de la fuerza y la distribución de los campos magnéticos en una muestra y mostrar la estructura de dominio magnético.
Más información ›
Adquisición de imágenes de muestras calientes
El estudio de los materiales en condiciones reales suele implicar el trabajo a altas temperaturas. El comportamiento de los materiales cuando se recristalizan, derriten, deforman o reaccionan ante el calor se puede estudiar in situ con la microscopía electrónica de barrido o con las herramientas DualBeam.
Más información ›
SEM ambiental (ESEM)
El SEM ambiental permite que se adquieran imágenes de los materiales en su estado nativo. Esto es ideal para investigadores académicos e industriales que necesitan probar y analizar muestras húmedas, sucias, reactivas, con liberación de gases o que no son compatibles con el vacío.
Más información ›
Espectroscopía de pérdida de energía de electrones
La investigación en ciencias de los materiales se beneficia de la EELS de alta resolución para una amplia gama de aplicaciones analíticas. Esto incluye asignación elemental de alto rendimiento, alta relación señal-ruido, así como sondeo de estados de oxidación y fonones de superficie.
Más información ›
Corte transversal
El corte transversal proporciona una visión adicional, ya que descubre información de la subsuperficie. Los instrumentos DualBeam tienen columnas FIB para poder realizar el corte transversal con alta calidad. Con la automatización, se puede realizar el procesamiento de muestras de alto rendimiento sin supervisión.
Más información ›
Experimentación in situ
La observación directa y en tiempo real de los cambios microestructurales con microscopía electrónica es necesaria para comprender los principios subyacentes de los procesos dinámicos como la recristalización, el crecimiento del grano y la transformación de fases durante el calentamiento, refrigeración y humectación.
Más información ›
Análisis de partículas
El análisis de partículas juega un papel vital en la investigación de nanomateriales y el control de calidad. La resolución a escala nanométrica y la adquisición de imágenes superiores de microscopía electrónica se pueden combinar con software especializado para la rápida caracterización de polvos y partículas.
Más información ›
SIMS
El detector TOF-SIMS (espectrometría de masas de iones secundaria de tecnología de tiempo de vuelo) para microscopía electrónica de tecnología barrido de haz de iones (FIB-SEM) permite la caracterización analítica de alta resolución de todos los elementos de la tabla periódica, incluso a bajas concentraciones.
Más información ›
Análisis de escala múltiple
Los novedosos materiales se deben analizar a una resolución cada vez mayor, manteniendo el contexto más amplio de la muestra. El análisis de escala múltiple permite la correlación de varias herramientas y modalidades de obtención de imágenes, tales como microTC de rayos X, DualBeam, PFIB láser, SEM y TEM.
Más información ›
Espectroscopía de fotoelectrones de rayos X
La espectroscopía de fotoelectrones de rayos X (XPS) permite el análisis de superficie, proporcionando la composición elemental, así como el estado químico y electrónico de los 10 nm principales de un material. Con la realización de perfiles de profundidad, el análisis XPS se extiende a la visión de composición de las capas.
Más información ›
Flujo de trabajo de partículas automatizado
El flujo de trabajo de nanopartículas automatizado (APW) es un flujo de trabajo de microscopio electrónico de transmisión para el análisis de nanopartículas que proporciona adquisición de imágenes de área extensa y de alta resolución, además de adquisición de datos en nanoescala, todo ello con un procesamiento sobre la marcha.
Más información ›
Products
Hoja de estilo para tarjetas originales instrumentos
Spectra 300
  • Información química y estructural de máxima resolución a nivel atómico
  • Intervalo de alta tensión flexible de 30-300 kV
  • Sistema de condensador de tres lentes
Spectra 200
  • Adquisición de imágenes de alta resolución y contraste para tensiones de aceleración de 30-200 kV
  • Lente de objetivo simétrico S-TWIN/X-TWIN con diseño de pieza polar de holgura amplia de 5,4 mm
  • Resolución de imágenes STEM subangstrom de 60 kV-200 kV
Talos F200C TEM
  • Análisis EDS flexible que revela información química
  • Adquisición de imágenes TEM y STEM de alto contraste y alta calidad
  • La cámara Ceta de 16 MP con sensor CMOS proporciona un amplio campo de visión y una rápida velocidad de lectura
Talos F200i TEM
  • Imágenes S/TEM y EDS precisas y de alta calidad
  • Disponible con tecnología EDS dual
  • Capacidades óptimas in situ
  • Amplio campo de adquisición de imágenes a grande a alta velocidad
Talos F200S TEM
  • Datos precisos de composición química
  • Adquisición de imágenes de alto rendimiento y análisis de composición preciso para microscopía dinámica
  • Cuenta con software Velox para una adquisición y análisis rápidos y fáciles de datos multimodales
Talos F200X TEM
  • Alta resolución/rendimiento en adquisición de imágenes STEM y análisis químicos
  • Añade soportes de muestras in situ para experimentos dinámicos
  • Cuenta con software Velox para una adquisición y análisis rápidos y fáciles de datos multimodales
Helios Hydra DualBeam
  • 4 especies de iones rápidos conmutables (Xe, Ar, O, N) para el procesamiento optimizado de PFIB de la gama más amplia de materiales
  • Preparación de muestras de TEM sin GA
  • Adquisición de imágenes SEM de alta resolución
Helios 5 PFIB DualBeam
  • Preparación de muestras STEM y TEM sin galio
  • Subsuperficie multimodal e información 3D
  • Columna FIB con plasma xenón de 2,5 μA de última generación
Helios 5 DualBeam
  • Preparación de muestras de TEM ultrafina, de alta calidad y completamente automatizada
  • Subsuperficie de alto rendimiento, alta resolución y caracterización en 3D
  • Capacidades de rápida creación de prototipos a nanoescala
Scios 2 DualBeam
  • Soporte completo de muestras magnéticas y no conductoras
  • Subsuperficie de alto rendimiento y caracterización en 3D
  • Facilidad de uso y capacidades de automatización avanzadas
Verios 5 XHR SEM
  • SEM monocroma para resolución de subnanómetros sobre el rango de energía completo de 1 keV a 30 keV
  • Fácil acceso a la energía de recepción de haz, a un nivel tan bajo como 20 eV
  • Excelente estabilidad con fase piezoeléctrica como estándar
Quattro ESEM
  • Tecnología de obtención de imágenes por microscopía electrónica de barrido (SEM) con cañón de emisión de campo (FEG) de alta resolución con versatilidad absoluta y capacidad medioambiental única (ESEM)
  • Consulte la información completa de todas las muestras con obtención de imágenes SE y BSE en cada modo de funcionamiento
Apreo 2 SEM
  • SEM de alto rendimiento para resolución de subnanómetro o nanómetro completo
  • Detector de retrodispersión de T1 incorporado en la columna para contraste de materiales sensibles de velocidad TV
  • Excelente rendimiento en distancias de trabajo largas (10 mm)
VolumeScope 2 SEM
  • Datos 3D isotrópicos de grandes volúmenes
  • Alto contraste y resolución en los modos de vacío alto y bajo
  • Un solo movimiento entre el uso de SEM normal y la adquisición de imágenes de cara de bloques en serie
SEM de escritorio Phenom Pro
  • SEM de escritorio de alto rendimiento
  • Resolución de <8 nm (SE) y <10 nm (BSE); aumento de hasta 150.000x
  • Detector SE opcional
SEM de escritorio Phenom ProX
  • SEM de escritorio de alto rendimiento con detector EDS integrado
  • Resolución de <8 nm (SE) y <10 nm (BSE); aumento de hasta 150.000x
  • Detector SE opcional
SEM de escritorio Phenom XL G2
  • Para muestras grandes (100 x 100 mm) e ideal para automatización
  • Resolución de <10 nm y aumento de hasta 200.000x; tensión de aceleración desde 4,8 kV hasta 20 kV
  • Detector de EDS y BSE, totalmente integrado, opcional
SEM de escritorio Phenom Pharos
  • Fuente FEG con un rango de tensión de aceleración desde 2 hasta 15 kV
  • Resolución de <2,5 nm (SE) y <4,0 nm (BSE) @ 15 kV; aumento de hasta 1.000.000x
  • Detector de EDS y SE, totalmente integrado, opcional
SEM de escritorio Phenom Pure
  • SEM de escritorio de nivel básico
  • Resolución de <25 nm; aumento de hasta 65.000x
  • Fuente CeB6 de larga duración
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Software Auto Slice and View 4.0
  • Cortes en secciones en serie automatizadas para DualBeam
  • Adquisición de datos multimodales (SEM, EDS, EBSD)
  • Capacidades de edición sobre la marcha
  • Ubicación del corte en los bordes
Software Avizo
  • Compatibilidad con varios datos/varias vistas, multicanal, series temporales, datos de gran tamaño
  • Registro automático avanzado multimodo 2D/3D
  • Algoritmos de reducción de artefactos
Software Inspect 3D
  • Filtros y herramientas de tratamiento de imágenes para correlación cruzada
  • Seguimiento de funciones para alineación de imágenes
  • Técnica de reconstrucción algebraica para la comparación de proyección iterativa
Software Maps
  • Adquiera imágenes de alta resolución en grandes áreas
  • Encuentre fácilmente áreas de interés
  • Automatice el proceso de adquisición de imágenes
  • Correlacione datos de diferentes fuentes
Reconstrucción en 3D
  • Interfaz de usuario intuitiva, uso máximo
  • Interfaz de usuario totalmente automatizada e intuitiva
  • Basada en la tecnología 'shape from shading' (forma a través de sombreados), sin inclinación de etapas necesaria
PoroMetric
  • Correlaciona las características de los poros como el área, la relación de aspecto, el eje principal y el secundario
  • Adquiera imágenes directamente desde el SEM de escritorio
  • Datos estadísticos con imágenes de alta calidad
ParticleMetric
  • Software integrado en ProSuite para análisis en línea y fuera de línea
  • Funciones de correlación de partículas como diámetro, circularidad, relación de aspecto y convexidad
  • Creación de conjuntos de datos de imágenes con el mapeo de imágenes automatizado
Elemental Mapping
  • Información rápida y fiable sobre la distribución de elementos de la muestra o la línea seleccionada
  • Resultados fácilmente exportados y comunicados
FiberMetric
  • Ahorre tiempo mediante mediciones automáticas
  • Recopilación rápida y automática de todos los datos estadísticos
  • Visualización y medición de nanofibras y microfibras con exactitud incomparable
μHeater
  • Solución de calentamiento ultrarrápido para adquisición de imágenes de alta resolución in situ
  • Totalmente integrada
  • Temperaturas de hasta 1200 °C
Soporte de muestras estándar
  • Etapa compacta que permite el análisis de muestras de hasta 100 mm x 100 mm
  • Se puede ampliar con 3 tipos de resina o insertos de montaje metalúrgicos
  • Se utiliza con el SEM de escritorio Phenom
Soporte de muestras eucéntricas
  • Rotación en dos dimensiones e inclinación eucéntrica en un SEM de escritorio
  • Tiempo rápido de obtención de imágenes con carga de muestras en < 1 minuto
  • Módulo de visualización de muestras 3D en tiempo real
Soporte de muestras de tracción
  • Determinación de la calidad del lote
  • Determinación de la uniformidad en la fabricación
  • Ayuda al proceso de diseño
Insertos de montaje de resina
  • Concepto exclusivo del soporte de muestras
  • Disponible en 3 modelos compatibles con muestras de tamaño estándar de 25 mm (~1 pulgada), 32 mm (aproximadamente 1⁄4 pulgada) y 40 mm (aproximadamente 1⁄4 pulgada) de diámetro