Tiling Analysis Software—version 1.1
 Download installer for Windows 64 bit OS | Doc
Version 1.1 includes the following changes:
  • Option to write either CHP or BAR file: Specify option in the Export tab of the Default Properties dialog.
  • Added the scaling target intensity to the file properties function.
  • Enrichment analysis: This analysis performs a simulation to assess the likelihood that the observed overlap is different from the expected overlap with no enrichment.
  • Interval overlap report: This report provides the number of intervals and bases from a BED file that are within a user-specified distance of intervals from a reference BED file.
  • PM only analysis of PM/MM arrays.
For Research Use Only. Not for use in diagnostic procedures.