Search
Flow cytometry and fluorescent-activated cell sorting (FACS) heavily rely on the user working with intact cells. Flow researchers must balance removal of cell debris and unwanted cells while preserving the integrity of cells-of-interest. Dynabeads FlowComp positive cell isolation kits positively isolate cells of interest directly followed by cell release and removal of magnetic beads. No sample preparation (e.g., density gradient centrifugation or lysis of red blood cells) is required, which helps eliminate the risk of contaminating and incubating your cells with iron, dextran, or other potentially harmful chemicals that are present in lysis buffers.
Get flow-ready cells in just three steps:
Explore flow cytometry reagents
Explore flow cytometry antibodies
Dynabeads magnetic beads combine with T cell markers to positively isolate specific T cell populations of interest. FlowComp positive isolation kits are available against four different human T cell markers.
Cells isolated with Dynabeads FlowComp kits keep their functional characteristics and are directly suitable for flow cytometry and any downstream cell-based assay.
View FlowComp protocol for isolation from human whole blood
Order Dynabeads FlowComp kits for human T cell markers
Dynabeads magnetic beads combine with T cell markers to positively isolate your specific T cell populations of interest for your mouse studies. FlowComp positive isolation kits are available against four different mouse T cell markers.
Cells isolated with Dynabeads FlowComp kits are significantly purer and more viable than isolation with a column-based method (Figure 1). These cells keep their functional characteristics and are suitable to be used directly for flow cytometry and any downstream cell-based assay.
Combine Dynabeads magnetic beads with your antibody of choice to isolate any cell type. The Dynabeads FlowComp Flexi Kit has a wide range of species reactivity. Starting samples are compatible with the build-your-own kit:
![]() | Compatible antibodies with FlowComp Dynabeads human CD4 positive isolation kit |
| Conjugate | Recommended antibody |
|---|---|
| Alexa Fluor 405 | CD4, Mouse anti-Human, (Alexa Fluor 405) |
| Alexa Fluor 488 | CD4, Mouse anti-Human, (Alexa Fluor 488) |
| Alexa Fluor 700 | CD4, Mouse anti-Human, (Alexa Fluor 700) |
| APC | CD4, Mouse anti-Human, (APC) |
| APC-Alexa Fluor 750 | CD4, Mouse anti-Human, (APC-Alexa Fluor 750) |
| APC-Cy5.5 | CD4, Mouse anti-Human, (APC-Cy5.5) |
| FITC | CD4, Mouse anti-Human, (FITC) |
| FITC | CD4, Mouse anti-Human, (FITC) |
| Pacific Blue | CD4, Mouse anti-Human, (Pacific Blue) |
| Pacific Orange | CD4, Mouse anti-Human, (Pacific Orange) |
| PE | CD4, Mouse anti-Human, (PE) |
| PE | CD4, Mouse anti-Human, (PE) |
| PE-Alexa Fluor 610 | CD4, Mouse anti-Human, (PE-Alexa Fluor 610) |
| PE-Alexa Fluor 700 | CD4, Mouse anti-Human, (PE-Alexa Fluor 700) |
| PE-Cy5.5 | CD4, Mouse anti-Human, (PE-Cy5.5) |
| PE-Cy7 | CD4, Mouse anti-Human, (PE-Cy7) |
| PE-Texas Red | CD4, Mouse anti-Human, (PE-TR) |
| TRI-COLOR | CD4, Mouse anti-Human, (TRI-COLOR) |
![]() | Compatible antibodies with FlowComp Dynabeads mouse CD4 positive isolation kit |
| Conjugate | Recommended antibody |
|---|---|
| Alexa Fluor 405 | CD4, Rat anti-Mouse, (Alexa Fluor 405) |
| Alexa Fluor 488 | CD4, Rat anti-Mouse, (Alexa Fluor 488) |
| APC | CD4, Rat anti-Mouse, (APC) |
| APC-Cy5.5 | CD4, Rat anti-Mouse, (APC-Cy5.5) |
| FITC | CD4, Rat anti-Mouse, (FITC) |
| Pacific Blue | CD4, Rat anti-Mouse, (Pacific Blue) |
| Pacific Orange | CD4, Rat anti-Mouse, (Pacific Orange) |
| PE | CD4, Rat anti-Mouse, (PE) |
| PE-Alexa Fluor 610 | CD4, Rat anti-Mouse, (PE-eFluor 610) |
| PE-Texas Red | CD4, Rat anti-Mouse, (PE-Texas Red) |
| TRI-COLOR | CD4, Rat anti-Mouse, (TRI-COLOR) |
![]() | Compatible antibodies with FlowComp Dynabeads human CD8 positive isolation kit |
| Conjugate | Recommended antibody |
|---|---|
| Alexa Fluor 405 | CD8, Mouse anti-Human, (Alexa Fluor 405) |
| Alexa Fluor 647 | CD8, Mouse anti-Human, (Alexa Fluor 647) |
| Alexa Fluor 700 | CD8, Mouse anti-Human, (Alexa Fluor 700) |
| APC | CD8, Mouse anti-Human, (APC) |
| APC-Alexa Fluor 750 | CD8, Mouse anti-Human, (APC-Alexa Fluor 750) |
| FITC | CD8, Mouse anti-Human, (FITC) |
| FITC | CD8, Mouse anti-Human, (FITC) |
| Pacific Blue | CD8, Mouse anti-Human, (Pacific Blue) |
| Pacific Orange | CD8, Mouse anti-Human, (Pacific Orange) |
| PE | CD8, Mouse anti-Human, (PE) |
| PE | CD8, Mouse anti-Human, (PE) |
| PE-Alexa Fluor 610 | CD8, Mouse anti-Human, (PE-Alexa Fluor 610) |
| PE-Cy5.5 | CD8, Mouse anti-Human, (PE-Cy5.5) |
| PE-Cy7 | CD8, Mouse anti-Human, (PE-Cy7) |
| PE-Texas Red | CD8, Mouse anti-Human, (PE-Texas Red) |
| TRI-COLOR | CD8, Mouse anti-Human, (TRI-COLOR) |
For information about tested clones from other suppliers than Thermo Fisher, please contact technical support.