Advanced FTIR technology for industrial gas analysis
Monitor and quantify process gases and emissions with confidence through sensitive, real-time analysis
Thermo Scientific™ MAX-iR™ FTIR Gas Analyzers have been engineered to meet the most demanding standards for in-line process, batch sampling, gas purity/certification, emissions testing, and ambient air monitoring. Powered by Thermo Scientific StarBoost™ Optical Enhancement Technology, the OE-FTIR MAX-iR Analyzer achieves single-digit ppb detection limits—and even mid-ppt performance in ultra-high-purity bulk gas applications.
With ultra-fast analysis and ultra-low detection limits, the MAX-iR FTIR Gas Analyzers provide an alternative to traditional techniques such as gas chromatography (GC) that can be slower, and more costly.
Instrument features
Instrument models
Find the right analyzer for your needs. 
Applications
Explore the applications where MAX FTIR gas analyzers are utilized.
Technology
Discover the advanced engineering that powers the MAX-iR FTIR gas analyzers.
Service and support
Explore service and support offerings.
FAQs
Get answers to the most frequently asked questions.
Resources
Download videos, brochures, app notes, and other resources from our e-library.
Service and product availability may vary by country and are subjected to varying regulatory requirements. Please contact your local sales representative for availability.