Search
Ultra-low and cryogenic freezers are designed to store samples rather than freeze them. Our blast freezers and controlled-rate freezers have been designed to properly prepare samples before the important storage phase, thus reducing stress on the storage freezer.
Thermo Scientific™ XBF40D blast freezers are designed to accommodate staggering loads and a variety of vessels such as carboys, bottles, racks with vials and more. Unlike previous generation blast freezers, the XBF40D is a plug and play requiring no customized installation and no remote compressors.
Thermo Scientific™ CryoMed™ controlled-rate freezers are designed as a complete, one-piece, benchtop freezing system. Modify six standard preset and ten user-defined freezing profiles for custom protocols to meet a variety of application needs.
Thermo Scientific™ Mr. Frosty™ is a simple to use system, designed to achieve a rate of cooling very close to -1°C per minute. Mr. Frosty accommodates tubes from 1 to 5mL.