Dionex Inuvion Core IC System: Simple, intuitive ion analysis
Perform reliable, basic ion analysis while maintaining low operating costs with the Thermo Scientific Dionex Inuvion Core IC System. This instrument provides all the benefits of chemical suppression and high-pressure operation, with the ability to upgrade features to perform additional applications as your needs and budget allow. Built on the latest innovative technology, the Dionex Inuvion Core IC System is designed to maximize productivity and uptime, so you can be sure your ion analyses are producing high-quality, reliable results.
Highlights
Flexible, upgradeable system
As a part of the Dionex Inuvion platform, this instrument can be easily upgraded from the Dionex Inuvion Core to the Dionex Inuvion (with or without Reagent-Free IC capabilities) as your analysis needs change. User-installable accessories can be swapped in and out of the instrument in minutes for extended application capabilities.
Maximum productivity and performance
The Dionex Inuvion Core IC System is built with state-of-the-art pump technology and electronics for improved day-to-day reliability. System self-diagnostics also detect issues with hardware and consumables, giving you the confidence that it will be ready to run whenever you are.
Simple user experience
The smart, function-driven engineering of this ion chromatography system allows users to quickly and easily access all necessary components, connections and consumables without a stepstool or extra tools.
Fast and easy setup
With automated startup and shutdown routines, the Dionex Inuvion Core IC System is quickly ready to run samples with minimal delay or user intervention. Simple how-to videos built into the software reduce training time and simplify overall setup and operation.
High-pressure operation
The Dionex Inuvion Core IC System can operate at pressures up to 5,000 psi. This pressure allows the instrument to support a full range of high-performance 4 µm-particle-size Thermo Scientific Dionex IC columns for improved speed and quality of results.
Comprehensive support and service
Ensure your Dionex Inuvion Core IC system runs smoothly and efficiently with the Unity™ Lab Services IonCare™ service plan*. This comprehensive 5-year plan covers parts, labor, and travel expenses, as well as preventive maintenance visits in years 2 and 4.
*IonCare service plan is available only in North America, Europe, the Middle East, Africa, Asia Pacific, and Japan
Videos
System specifications
The table below provides an overview of the Dionex Inuvion Core IC System specifications. Not all specifications are listed. For a full list of the specifications, download the Dionex Inuvion Core IC System specifications sheet.
Specification
Dionex Inuvion Core IC System
Analytical pump and fluidics
Type
Serial dual-reciprocating pistons, microprocessor-controlled constant stroke, variable speed
Construction
Chemically inert, metal-free PEEK pump heads and flow paths compatible with aqueous eluents of pH 0–14 and reversed-phase solvents
Pump operating pressure
0–35 MPa (0–5000 psi)
Flow rate range
0.00–5.00 mL/min in 0.01 mL/min increments
Flow precision
<0.1%, typically
Flow accuracy
<0.1%, typically
Pressure ripple
<1%
Eluent on-off valve
Standard
Leak sensor
Optical, standard
Piston seal wash (optional)
Pump head wash can be operated in continuous or intermittent mode when connected to rinse solution supply
Pressure alarm limits
Upper and lower limit pressure alarms can be set
Eluent bottles
Standard 2 L polypropylene bottle; allows various sizes
Eluent bottle pressure regulator (optional)
Supported by digital controlled regulator with display
Injection valve
6-port, 2-position Rheodyne valve, electronically activated
Columns supported
2, 3, 4, and 5 mm ID; maximum length 250 mm analytical column with 50 mm guard column
Column heater (optional)
Operating temperature range
10 to 60 °C (50 to 140 °F); settable within the software; minimum working range is 5 °C above ambient temperature
Temperature accuracy
±0.5 °C at sensor, at calibration points (35, 45 °C)
Suppressors and control
Chemical suppression
2 mm and 4 mm anion and cation suppressor types
Carbonic acid removal for anions
Thermo Scientific Dionex CRD 200 Carbonate Removal Device for use with hydroxide eluents or Dionex CRD 300 for use with carbonate eluents
Non-suppressed chromatography
Yes, supported
Suppressor wear parts
Optional regenerant pump for chemical suppressor
Dynamic suppression capacity
Anions:
  • Thermo Scientific Dionex ACRS 500 (4 mm): 150 μeq/min
  • Thermo Scientific Dionex ACRS 500 (2 mm): 75 μeq/min
Cations:
  • Thermo Scientific Dionex CCRS 500 (4 mm): 75 μeq/min
  • Thermo Scientific Dionex CCRS 500 (2 mm): 37.5 μeq/min
Void volumes
  • <50 μL for 4mm Thermo Scientific Dionex CRS 500 suppressors
  • <15 μL for 2mm Dionex CRS 500 suppressors
Conductivity detector electronics and flow cell
Type
Microprocessor-controlled digital signal processor
Cell drive
128 kHz square wave
Linearity
r2≥0.999%
Resolution
0.002 nS/cm
Full-scale output ranges
Digital input signal range 0-18,000 μS/cm, with auto-ranging; analog output signal range 0-18,000 μS/cm
Temperature compensation
Variable, default set at 1.7%/°C at cell temperature
Temperature range
Ambient +7 °C, 15 to 60 °C
Cell electrodes
Passivated 316 stainless steel; compatible with MSA
Cell body
Chemically inert polymeric material
Cell volume
<1 µL
Heat exchanger
Inert, tortuous path for low axial dispersion
Maximum cell operating pressure
10 MPa (1,500 psi)
Data filter
Rise times from 0 to 10 s, Data Collection Rate 1 to 100 Hz, user selectable
Autosampler
Automation using autosampler
For more information about available autosamplers, visit IC Autosamplers
System software
Chromeleon CDS, requires Microsoft® Windows® 10 or 11
Physical specifications
Dimensions (h × w × d)
66.1 x 29.2 x 43.2 cm (26.0 x 11.5 x 17.0 in.)
Weight
15.3 kg (33.7 lb)
Power requirements
100–240 V AC, 50–60 Hz autoranging
Operating temperature
4–40 °C (40–104 °F)
Operating humidity range
20-80% relative, non-condensing
Control modes
Full control through Chromeleon CDS software; alternative control through TTL or relay closures; one relay output, two TTL outputs, two assignable TTL inputs
USB communication protocol
One USB input; three USB outputs
Ordering information
Instruments
Optional accessories
For Research Use Only. Not for use in diagnostic procedures.