Designed to meet EPA requirements 40 CFR 75 and 40 CFR 60 
Support your emissions and gas monitoring operations with our line of Thermo Scientific emissions monitoring probes and controllers. Our probes and controllers are built to provide accurate and concise readings in the toughest environments, whether you need information about source gas, industrial emissions, concentration, or mass-emission rates.
Low-cost installation, maintenance and operation
Handle the toughest stack environments with our flow and low flow dilution probes.  These probes are modular, external, and designed for wet- or dry-basis measurement and offer significant advantages over typical in-stack dilution probes. Our dilution extractive technology in combination with the thermo-electric cooler provides accurate and concise readings. Conduct wet- or dry-basis measurements with our full extractive probe that is highly durable and easily mountable to the stack or duct. Our diluting stack sampler performs the four critical functions of sample delivery, course filtering, fine filtering, and sample dilution.
Thermo Scientific PRO903 Dilution Probe
Thermo Scientific Model PRO902C Low Flow Dilution Probe
Thermo Scientific PRO2001WHP Dilution Extractive Probe
Thermo Scientific Model PRO3000 Full Extractive Probe
Thermo Scientific EPM.300 Series Diluting Stack Sampler
Service and product availability may vary by country and are subject to varying regulatory requirements. Please contact your local sales representative for availability.