Search
Ion channels provide exciting opportunities for drug development targeting a diverse array of therapeutic areas including neurological, cardiovascular and pulmonary diseases, pain, and inflammation. To address the most challenging ion channel targets in any desired cell background, researchers need an assortment of optimized assays for their discovery efforts. Our product offering for cell-based ion channel assays is designed for sensitivity, ease-of-use, and assay flexibility.
| Ion | Assay Technology | Description |
|---|---|---|
| Potassium | FluxOR | Functional, HTS-validated universal potassium assay |
| BacMam Ion Channel Targets | BacMam-mediated delivery of potassium ion channels including hERG | |
| Calcium | Fluo-4 Direct | Add and read, homogenous fluorescent calcium readout validated for HTS |
| Chloride | Premo Halide | Fluorescent protein-based chloride biosensor protein |
| Membrane Potential | Voltage Sensor Probes | FRET-based, ratiometric assessment of membrane potential |
For Research Use Only. Not for use in diagnostic procedures.