Search
Nalgene cleaning supplies are build to last in a busy lab with rugged construction from plastic materials designed for repeated chemical exposure. From sink to autoclave; pans, baskets, buckets, and pipet washing equipment, we've got your toughest lab cleaning jobs covered.
Nalgene buckets, pails and storage tubs come in a variety of configurations, but all have one thing in common - excellent chemical compatibility with a wide range of laboratory chemicals and production materials.
Nalgene Pans, Trays and Baskets are made rugged with the rigors of lab life in mind. Choose from an assortment of polypropylene pans, trays and baskets for autoclaving and sterilizing labware and instruments.
Soak and wash reusable pipets with the Nalgene pipet cleaning system. Choose the jar, basket and washer set that best fits your pipets, or purchase the three pieces separately and make your own configuration.
Thoroughly clean all plastic labware, especially polycarbonate, with Thermo Scientific Nalgene L900 Liquid Detergent. This environmentally friendly formula will not cause crazing, stress cracking or clouding of plastic when used as directed.
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.