Search
SoftFit-L pipette tips are specifically designed to fit Rainin LTS- style pipettes. The cylindrical collar and positive-stop design enable an exact fit, reducing attachment and ejection forces. With five essential tip volumes available, you can easily select the appropriate tips for your specific application. SoftFit-L tips offer the same level of accuracy, precision, and comfort you expect from Rainin LTS pipette tips.
Tip Type |
Benefits |
Recommended Use |
|---|---|---|
|
|
Blocks aerosols from contaminating the pipette and prevents cross-contamination of samples |
|
|
|
Economic, high-quality tips |
|
|
|
Hydrophobic surface maximizes sample delivery and conservation of expensive reagents |
|
Packaging Type |
Features and Benefits |
|
|---|---|---|
|
|
![]() |
|
|
|
|
|
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.