Search
| Feature | Nuclease Protection Assays | Northern Blotting | RT-PCR | In Situ Hybridization |
|---|---|---|---|---|
| Relative and Absolute Quantitation | Yes | Yes | Yes | Yes |
| Detection Limit (Copies of mRNA) | 4,000-5,000 | 10,000 | 1 (theoretical) | Medium |
| Level of Optimization Required | Medium | Medium | High | High |
| Sample Size | 100 µg max | 30 µg max | Not critical | Tissue sections |
| Detect Multiple Transcripts | Yes, in same tube | Yes, generally requires stripping and reprobing | Yes, in same tube | No |
| Localizes mRNA Expression within Tissues/Cells | No | No | No | Yes |
| Mapping Studies | Yes | No | No | No |
| Resolve Comigrating mRNAs | Yes | No | Yes | No |
| Sizing mRNAs | No | Yes | No | No |
| Detection of Alternatively Spliced Transcripts | No | Yes | No | No |
| Distinguish Between Members of Multi-Gene Families | Yes | Only if they are different sizes | Only if they are characterized | No |
| Tolerates Partially Degraded RNA | Yes | No | Yes | Yes |
| Advantages |