Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • E-cadherin Antibodies

          Invitrogen

          E-cadherin Monoclonal Antibody (ECCD-2)

          99 Published Figures
          382 References
          View all (166) E-cadherin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite E-cadherin Monoclonal Antibody (ECCD-2)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (99)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          E-cadherin Antibody (13-1900) in ICC/IF

          The Journal of physiology 2019 - Published figure using E-cadherin monoclonal antibody (Product # 13-1900) in Immunocytochemistry {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          E-cadherin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunocytochemistry (ICC/IF)
          E-cadherin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          E-cadherin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          E-cadherin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          E-cadherin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          E-cadherin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          E-cadherin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          E-cadherin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          E-cadherin Antibody in Western Blot, Immunohistochemistry (Paraffin) (WB, IHC (P))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)
          E-cadherin Antibody in Immunohistochemistry (IHC)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          13-1900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          10 µg/mL
          View 43 publications 43 publications

          Immunohistochemistry (IHC)

          -
          View 153 publications 153 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 20 publications 20 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          10 µg/mL
          View 20 publications 20 publications

          Immunocytochemistry (ICC/IF)

          -
          View 94 publications 94 publications

          Flow Cytometry (Flow)

          -
          View 12 publications 12 publications

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 34 publications 34 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Avian, Dog, Fish, Hamster, Human, Mouse, Pig, Rat, Tag

          Host/Isotype

          Rat / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          ECCD-2

          Immunogen

          Mouse liver E-cadherin fragment.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          2.0 mg/mL once reconstituted

          Amount

          100 µg

          Purification

          Ion-exchange chromatography

          Storage buffer

          TBS, pH 7.5, with 1% BSA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533005

          Product Specific Information

          Source: Monoclonal antibody was obtained by fusing the P3-X63-Ag8 mouse myeloma cell line with spleen cells of Donryu rat after immunization with mouse teratocarcinoma and mouse liver E-cadherin fragment.

          Specificity: This antibody specifically reacts with mouse E-cadherin. This antibody does not inhibit E-cadherin-dependent cell-cell contact.

          Cross reactivity: This antibody reacts with human and dog E-cadherin. This antibody does not react with bovine or chicken E-cadherin.

          Western blot can be performed under reducing or non-reducing conditions.

          Reconstitution and dilution: dissolve the lyophilized antibody in 50 µl of distilled water to a final concentration 2.0 mg/mL. This solution can be used as a stock solution. If dilution is necessary for your application, dilute the stock solution in 500 µL or more of the following dilution solution: 20mM TBS (pH 7.5), 10mM CaCl2, 1.0% BSA, 0.1% NaN3. Note that TBS should be used for dilution, as PBS causes precipitation.

          Storage: When stored at 4°C, 0.1% sodium azide should be added as a preservative.Store the antibody at a minimum concentration of 2.0 mg/mL; a lower antibody concentration may result in decreased stability.The stock solution (2.0mg/mL) should be stored in aliquots at -20°C for 1year, or stored at 4°C for 6 months after adding 0.1% sodium azide. Avoid repeated freeze-thaw cycles. Diluted antibody should not be stored.

          Target Information

          E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cadherin e; cadherin-E; E-cadherin; Ecadherin; epithelial cadherin; unnamed protein product; uvomorulin

          View more View less

          Gene Aliases: ARC-1; E-cad; Ecad; L-CAM; Um; UVO

          View more View less

          UniProt ID: (Mouse) P09803

          View more View less

          Entrez Gene ID: (Mouse) 12550

          View more View less

          Function(s)
          calcium ion binding protein binding beta-catenin binding cytoskeletal protein binding protein phosphatase binding protein domain specific binding ankyrin binding GTPase activating protein binding identical protein binding alpha-catenin binding gamma-catenin binding cadherin binding cell adhesion molecule binding binding, bridging cell adhesion mediator activity protein binding involved in cell-cell adhesion protein tyrosine kinase binding
          Process(es)
          cell morphogenesis in utero embryonic development trophectodermal cell differentiation desmosome assembly epithelial cell morphogenesis cell-cell junction assembly cell adhesion homophilic cell adhesion via plasma membrane adhesion molecules synapse assembly embryo implantation sensory perception of sound regulation of gene expression negative regulation of protein processing calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules cell migration negative regulation of cell-cell adhesion positive regulation of cell-cell adhesion actin cytoskeleton organization negative regulation of cell migration negative regulation of axon extension regulation of protein localization cell junction assembly adherens junction organization uterine epithelium development positive regulation of protein import into nucleus cell-cell adhesion mediated by cadherin positive regulation of transcription, DNA-templated decidualization negative regulation of epithelial cell proliferation intestinal epithelial cell development salivary gland cavitation regulation of branching involved in salivary gland morphogenesis establishment of skin barrier bicellular tight junction assembly cellular response to amino acid stimulus cellular response to lithium ion cellular response to indole-3-methanol protein localization to plasma membrane negative regulation of canonical Wnt signaling pathway cochlea development cell-cell adhesion regulation of protein catabolic process at postsynapse, modulating synaptic transmission negative regulation of protein localization to plasma membrane positive regulation of cellular protein localization regulation of protein localization to cell surface regulation of neuron migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch