Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TER-119 Antibodies

          Invitrogen

          TER-119 Monoclonal Antibody (TER-119), Functional Grade, eBioscience™

          57 Published Figures
          50 References
          View all (39) TER-119 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TER-119 Monoclonal Antibody (TER-119), Functional Grade, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (57)
          TER-119 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          TER-119 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 57

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TER-119 Antibody (16-5921-85) in IHC

          The Journal of clinical investigation 2018 - Published figure using TER-119 monoclonal antibody (Product # 16-5921-85) in Immunohistochemistry {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Immunohistochemistry (IHC)
          TER-119 Antibody in Western Blot (WB)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)
          TER-119 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          16-5921-85

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 42 publications 42 publications

          Functional Assay (Functional)

          Assay-Dependent
          -

          Control (Ctrl)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Fish, Human, Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          TER-119

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Unconjugated
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 480
          • eFluor 450
          • eFluor 506
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          500 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_469153

          Product Specific Information

          Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin.

          Applications Reported: The TER-119 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The TER-119 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ly-76; Ly76; TER119

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0