DMS (dimethyl suberimidate)
Thermo Scientific™

DMS (dimethyl suberimidate)

Thermo Scientific Pierce DMS is dimethyl suberimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end ofRead more
Have Questions?
Catalog NumberQuantity
207001 g
Catalog number 20700
Price (USD)
204.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
204.00
Each
Thermo Scientific Pierce DMS is dimethyl suberimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of an 8-atom spacer arm.

Features of dimethyl suberimidate:

• Reactive groups: imidoester (both ends)
• Reactive towards: amino groups (primary amines)
• Longest of three similar imidoester crosslinkers, which include DMA, DMP and DMS
• Water-soluble
• Imidoesters react with amines at alkaline pH values (pH 8-10) to form amidine bond
• Amidine bond retains net charge character of protein primary amine to which it reacts
• Reversible at high pH values
• Tool for study of quarternary structure of proteins

Product References
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
DMA (dimethyl adipimidate)
DMP (dimethyl pimelimidate)
DTBP (Wang and Richard's Reagent)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityYes
DescriptionDMS
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)273.2
PEGylatedNo
Product LinePierce
Quantity1 g
Reactive MoietyImidoester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length11.0 Å
Water SolubleYes
Chemical ReactivityAmine-Amine
CleavableNo
Crosslinker TypeHomobifunctional
FormatStandard
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for DMS (dimethyl suberimidate)?

DMS (dimethyl suberimidate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (5)

Citations & References
Abstract
Discovery of lung cancer biomarkers by profiling the plasma proteome with monoclonal antibody libraries.
Authors:Guergova-Kuras M, Kurucz I, Hempel W, Tardieu N, Kádas J, Malderez-Bloes C, Jullien A, Kieffer Y, Hincapie M, Guttman A, Csánky E, Dezso B, Karger BL, Takács L
Journal:Mol Cell Proteomics
PubMed ID:21947365
'A challenge in the treatment of lung cancer is the lack of early diagnostics. Here, we describe the application of monoclonal antibody proteomics for discovery of a panel of biomarkers for early detection (stage I) of non-small cell lung cancer (NSCLC). We produced large monoclonal antibody libraries directed against the ... More
The structural characterization of a prophage-encoded extracellular DNase from Streptococcus pyogenes.
Authors:Korczynska JE, Turkenburg JP, Taylor EJ
Journal:Nucleic Acids Res
PubMed ID:21948797
'The pathogenic bacterium Group A Streptococcus pyogenes produces several extracellular DNases that have been shown to facilitate invasive infection by evading the human host immune system. DNases degrade the chromatin in neutrophil extracellular traps, enabling the bacterium to evade neutrophil capture. Spd1 is a type I, nonspecific ßßa/metal-dependent nuclease from ... More
Outer membrane lipoprotein Lpp is Gram-negative bacterial cell surface receptor for cationic antimicrobial peptides.
Authors:Chang TW, Lin YM, Wang CF, Liao YD
Journal:J Biol Chem
PubMed ID:22084237
Antimicrobial peptides/proteins (AMPs) are important components of the host innate defense mechanisms. Here we demonstrate that the outer membrane lipoprotein, Lpp, of Enterobacteriaceae interacts with and promotes susceptibility to the bactericidal activities of AMPs. The oligomeric Lpp was specifically recognized by several cationic a-helical AMPs, including SMAP-29, CAP-18, and LL-37; ... More
Inhibition of RNase L and RNA-dependent protein kinase (PKR) by sunitinib impairs antiviral innate immunity.
Authors:Jha BK, Polyakova I, Kessler P, Dong B, Dickerman B, Sen GC, Silverman RH
Journal:J Biol Chem
PubMed ID:21636578
RNase L and RNA-dependent protein kinase (PKR) are effectors of the interferon antiviral response that share homology in their pseudokinase and protein kinase domains, respectively. Sunitinib is an orally available, ATP-competitive inhibitor of VEGF and PDGF receptors used clinically to suppress angiogenesis and tumor growth. Sunitinib also impacts IRE1, an ... More
Human mitochondrial DNA helicase TWINKLE is both an unwinding and annealing helicase.
Authors:Sen D, Nandakumar D, Tang GQ, Patel SS
Journal:J Biol Chem
PubMed ID:22383523
TWINKLE is a nucleus-encoded human mitochondrial (mt)DNA helicase. Point mutations in TWINKLE are associated with heritable neuromuscular diseases characterized by deletions in the mtDNA. To understand the biochemical basis of these diseases, it is important to define the roles of TWINKLE in mtDNA metabolism by studying its enzymatic activities. To ... More