Pierce™ Recombinant Protein L
Thermo Scientific™

Pierce™ Recombinant Protein L

Purified (unconjugated) Thermo Scientific Pierce Recombinant Protein L is useful as the basis for preparing various kinds of probes orRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
211891 mg
7767950 mg
Catalog number 21189
Price (USD)
125.65
Online exclusive
140.00
Save 14.35
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
125.65
Online exclusive
140.00
Save 14.35
Each
Purified (unconjugated) Thermo Scientific Pierce Recombinant Protein L is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of mouse and human antibodies in immunoassays and antibody purification protocols.

Features of Recombinant Protein L:

• Contains four immunoglobulin-binding domains per protein
• Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
• Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Properties of Recombinant Protein L:
• Source: E. coli
• Molecular Weight: ∼35,800 (Apparent MW by SDS-PAGE: 36,000)
• Form: salt-free powder
• A280 of 0.1% solution: ∼0.80
• Isoelectric point (pI): 4.8

Related Products
Pierce™ Recombinant Protein L, Biotinylated
Pierce™ Recombinant Protein L, Peroxidase Conjugated
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormPowder
Protein SubtypeProtein L
TargetAntibodies
Product LinePierce
Product TypeProtein L
Quantity1 mg
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C.