Pierce™ Recombinant Protein L, Peroxidase Conjugated
Thermo Scientific™

Pierce™ Recombinant Protein L, Peroxidase Conjugated

Use Thermo Scientific Pierce Recombinant Peroxidase Conjugated Protein L to probe and detect mouse and human antibodies containing kappa light詳細を見る
製品番号(カタログ番号)数量
324200.5 mg
製品番号(カタログ番号) 32420
価格(JPY)
54,100
0.5 mg
数量:
0.5 mg
一括またはカスタム形式をリクエストする
Use Thermo Scientific Pierce Recombinant Peroxidase Conjugated Protein L to probe and detect mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.

Features of Recombinant Protein L:

• Contains four immunoglobulin-binding domains per protein
• Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
• Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Properties of Recombinant Protein L:
• Source: E. coli
• Molecular Weight: ∼35,800 (Apparent MW by SDS-PAGE: 36,000)
• Form: salt-free powder
• A280 of 0.1% solution: ∼0.80
• Isoelectric point (pI): 4.8

Related Products
Pierce™ Recombinant Protein L
Pierce™ Recombinant Protein L, Biotinylated
For Research Use Only. Not for use in diagnostic procedures.
仕様
形状Powder
タンパク質サブタイプProtein L
ターゲットAntibodies
コンジュゲートHRP
製品ラインPierce
製品タイプProtein L, Peroxidase Conjugate
数量0.5 mg
Unit Size0.5 mg
組成および保存条件
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.