Pierce™ Recombinant Protein L, Biotinylated
Thermo Scientific™

Pierce™ Recombinant Protein L, Biotinylated

Use Thermo Scientific Pierce Biotinylated Recombinant Protein L to probe and detect mouse and human antibodies containing kappa light chainsRead more
Have Questions?
Catalog NumberQuantity
299970.5 mg
Catalog number 29997
Price (USD)
328.00
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
328.00
Each
Use Thermo Scientific Pierce Biotinylated Recombinant Protein L to probe and detect mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.

Features of Recombinant Protein L:

• Contains four immunoglobulin-binding domains per protein
• Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
• Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Properties of Recombinant Protein L:
• Source: E. coli
• Molecular Weight: ∼35,800 (Apparent MW by SDS-PAGE: 36,000)
• Form: salt-free powder
• A280 of 0.1% solution: ∼0.80
• Isoelectric point (pI): 4.8

Related Products
Pierce™ Recombinant Protein L
Pierce™ Recombinant Protein L, Peroxidase Conjugated
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormPowder
Protein SubtypeProtein L
TargetAntibodies
ConjugateBiotin
Product LinePierce
Product TypeProtein L, Biotinylated
Quantity0.5 mg
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C. Product is shipped at ambient temperature.