Product Image
Thermo Scientific™
EZ-Link™ Micro Sulfo-NHS-SS-Biotinylation Kit
The Thermo Scientific EZ-Link Sulfo NHS-SS Biotinylation Kit contains sufficient reagents for 8 biotinylation reactions (e.g., 0.05–0.2 mg antibody perRead more
Have Questions?
Catalog NumberQuantity
219458 reactions
Catalog number 21945
Price (MXN)
-
Quantity:
8 reactions
Request bulk or custom format
The Thermo Scientific EZ-Link Sulfo NHS-SS Biotinylation Kit contains sufficient reagents for 8 biotinylation reactions (e.g., 0.05–0.2 mg antibody per reaction). The EZ-Link Sulfo-NHS-SS-Biotin included in the kit is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.
Features of EZ-Link Sulfo-NHS-SS-Biotin:
• Protein labeling – biotinylate antibodies to facilitate immobilization, purification or detection
• Cell surface labeling – biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
• Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
• Cleavable—disulfide bond in spacer arm allows the biotin label to be removed using reducing agents such as DTT; only a small sulfhydryl group remains attached to the molecule
• Soluble – charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
• Medium length—spacer arm (total length added to target) is 24.3 angstroms; it consists of the native biotin valeric acid group extended by a 7-atom chain
Sulfo-NHS-SS-Biotin is a thiol-cleavable amine-reactive biotinylation reagent that contains an extended spacer arm to reduce steric hindrances associated with avidin binding. This reagent is water-soluble, enabling biotinylation to be performed in the absence of organic solvents such as DMSO or DMF for applications that cannot tolerate solvents or are complicated by their inclusion. The compound is particularly useful for labeling and purifying cell surface proteins, because (a) its sulfonate group prevents it from permeating cell membranes and (b) its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.
We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency, and performance for the intended research applications.
N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins (e.g., antibodies) typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide.
Varieties of biotin NHS-ester reagents differ in length, solubility, cell permeability and cleavability. Non-sulfonated NHS-biotins are cell permeable but must be dissolved in organic solvent such as DMSO or DMF. Sulfo-NHS biotins (and those with pegylated spacers) are directly water soluble but not membrane permeable. Varieties containing disulfide bonds can be cleaved using reducing agents, enabling the biotin group to be disconnected from the labeled protein.
Related Products
EZ-Link™ Sulfo-NHS-SS-Biotin
EZ-Link™ Sulfo NHS-SS Biotinylation Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionEZ-Link Micro Sulfo-NHS-SS-Biotinylation Kit
Labeling Scale100 μg
Product LineEZ-Link
Product TypeMicro Sulfo-NHS-SS-Biotinylation Kit
Quantity8 reactions
Labeling TargetAntibodies, Proteins
Label or DyeBiotin
SolubilityWater
Unit SizeEach
Contents & Storage
Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link Sulfo-NHS-SS-Biotin, 8 x 1 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns
Store biotin reagent at -20°C. Store remaining kit components at 4°C.
Frequently asked questions (FAQs)
Is there a cleavable analog of Sulfo-NHS-LC-Biotin?
Yes, we offer Sulfo-NHS-SS-Biotin (Cat. Nos. 21331, 21328). It has a disulfide bond in the spacer arm that can be cleaved with DTT or other reducing agents, thereby releasing the biotin tag from the previously labeled protein. This product is also available in kit format (Cat. Nos. 21945, 21445).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Which EZ-Link Micro Biotinylation Kit is best for my application?
All four Micro Biotinylation Kits will label proteins for use in many applications. Consider the following guidelines for choosing among the kits:
• Choose the Sulfo-NHS-Biotinylation Kit (Cat. No. 21925) when it is important to minimize the molecular mass contributed by the label.
• Choose the Sulfo-NHS-SS-Biotinylation Kit (Cat. No. 21945) only when the downstream application require cleaving the biotin tag with a reducing agent.
• Choose the NHS-PEG4-Biotinylation Kit (Cat. No. 21955) to maximize protein solubility and prevent protein aggregation during storage.
• Choose the Sulfo-NHS-LC-Biotinylation Kit (Cat. No. 21935) for routine purposes when a feature of one of the other reagents is not required. The longer spacer arm offers the possibility of optimizing detection experiments where steric hindrance of biotin binding is an important factor.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How long does it take to biotinylate a protein using the EZ-Link Micro Biotinylation kits?
The entire procedure can be completed in less than 3 hours.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What types of proteins can be labeled with the EZ-Link Micro Biotinylation kits?
Any molecule containing a primary amine can be labeled. Any protein that has been labeled using the standard biotinylation kits, including antibodies and peptides, also can be labeled with the micro kits.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Are the biotinylation reagents in the EZ-Link Micro Biotinylation Kits cleavable?
Only the sulfo-NHS-SS-Biotin is cleavable because it has a disulfide bond in its spacer arm, enabling labeled proteins to be cleaved from the biotin group by treatment with reducing agents.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.