Opti-MEM™ I Reduced Serum Medium
Gibco™

Opti-MEM™ I Reduced Serum Medium

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
319850701 x 500 mL
3198508810 x 500 mL
319850621 x 100 mL
Catalog number 31985070
Price (USD)
75.65
Online exclusive
83.50
Save 7.85
Each
Quantity:
1 x 500 mL
Price (USD)
75.65
Online exclusive
83.50
Save 7.85
Each

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.

This Opti-MEM™ I is modified as follows:

With: L-glutamine, Phenol Red

The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium

Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineSp2, AE-1, CHO, BHK-21, and HEK
Cell TypeMammalian Cells
Concentration1X
Culture EnvironmentCO2
For Use With (Application)Mammalian Cell Culture, Lipid-Based Transfection
FormatBottle
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco, Opti-MEM
Product TypeOpti-MEM I
Quantity1 x 500 mL
Shelf Life18 Months From Date of Manufacture
Shipping ConditionAmbient
ClassificationHuman Origin
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelReduced Serum
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo FBS
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture

Frequently asked questions (FAQs)

What should I use to dilute my RNAi duplexes and Lipofectamine RNAiMAX before complexing?

We recommend using Opti-MEM I Reduced Serum Medium (Cat. No. 31985062).

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Do you offer an alternative for Opti-MEM I Reduced Serum Medium (Cat. No. 31985070)?

Opti-MEM I Reduced Serum Medium, no phenol red (Cat. No. 11058021) would be the closest alternative. Both offerings contain L-glutamine. For more a stabilized L-glutamine option, Opti-MEM I Reduced Serum Medium, GlutaMAX Supplement (Cat. No. 51985034) is the recommended alternative. All Opti-MEM offerings have demonstrated performance for both reduced-serum cell culture and lipid-based transfection.

My OpitMEM Reduced Serum Medium (Cat. No. 31985070) was accidentally frozen. Can I still use it?

This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is there any other difference between Opti-MEM I Reduced Serum Medium and Opti-MEM I Reduced Serum Medium, no phenol red besides the presence/absence of phenol red?

The formulation of Opti-MEM I Reduced Serum Medium and that of Opti-MEM I Reduced Serum Medium, no phenol red is the same except for the presence of phenol red in the former medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Where can I find the osmolality for my lot of Opti-MEM I Reduced Serum Medium?

We do provide osmolality information on the certificate of analysis. All lots of Opti-MEM I Reduced Serum Medium will meet the osmolality specification of ≥280 to ≤320 mOsm/kg.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (10)

Citations & References
Abstract
Regulation of cortical dendrite development by Slit-Robo interactions.
Authors:Whitford Kristin L; Marillat Valérie; Stein Elke; Goodman Corey S; Tessier-Lavigne Marc; Chédotal Alain; Ghosh Anirvan;
Journal:Neuron
PubMed ID:11779471
Slit proteins have previously been shown to regulate axon guidance, branching, and neural migration. Here we report that, in addition to acting as a chemorepellant for cortical axons, Slit1 regulates dendritic development. Slit1 is expressed in the developing cortex, and exposure to Slit1 leads to increased dendritic growth and branching. ... More
Identification of a novel redox-sensitive gene, Id3, which mediates angiotensin II-induced cell growth.
Authors:Mueller Cornelius; Baudler Stephanie; Welzel Hilke; Böhm Michael; Nickenig Georg;
Journal:Circulation
PubMed ID:12021231
BACKGROUND: Reactive oxygen species, such as superoxide (O(2)(-)), are involved in the abnormal growth of various cell types. Angiotensin II (Ang II) is one of the most potent inducers of oxidative stress in the vasculature. The molecular events involved in Ang II-induced proliferation of vascular smooth muscle cells (VSMCs) are ... More
Non-viral-mediated gene transfer of OX40 ligand for tumor immunotherapy.
Authors:Rakitina OA,Kuzmich AI,Bezborodova OA,Kondratieva SA,Pleshkan VV,Zinovyeva MV,Didych DA,Sass AV,Snezhkov EV,Kostina MB,Koksharov MO,Alekseenko IV
Journal:Frontiers in immunology
PubMed ID:39007148
BACKGROUND: Immune checkpoint blockade (ICB) is rapidly becoming a standard of care in the treatment of many cancer types. However, the subset of patients who respond to this type of therapy is limited. Another way to promote antitumoral immunity is the use of immunostimulatory molecules, such as cytokines or T ... More
Effects of an indole derivative on cell proliferation, transfection, and alternative splicing in production of lentiviral vectors by transient co-transfection.
Authors:Mier NC,Roper DK
Journal:PloS one
PubMed ID:38833479
Lentiviral vectors derived from human immunodeficiency virus type I are widely used to deliver functional gene copies to mammalian cells for research and gene therapies. Post-transcriptional splicing of lentiviral vector transgene in transduced host and transfected producer cells presents barriers to widespread application of lentiviral vector-based therapies. The present study ... More
Aldose reductase mediates mitogenic signaling in vascular smooth muscle cells.
Authors: Ramana Kota V; Chandra Deepak; Srivastava Sanjay; Bhatnagar Aruni; Aggarwal Bharat B; Srivastava Satish K;
Journal:J Biol Chem
PubMed ID:12063254
'Abnormal vascular smooth muscle cell (VSMC) proliferation is a key feature of atherosclerosis and restenosis; however, the mechanisms regulating growth remain unclear. Herein we show that inhibition of the aldehyde-metabolizing enzyme aldose reductase (AR) inhibits NF-kappa B activation during restenosis of balloon-injured rat carotid arteries as well as VSMC proliferation ... More