Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Claudin 4 Antibodies

          Invitrogen

          Claudin 4 Monoclonal Antibody (3E2C1)

          Advanced Verification
          82 Published Figures
          300 References
          View all (19) Claudin 4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Claudin 4 Monoclonal Antibody (3E2C1)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (82)
          • Advanced Verification (1)
          Claudin 4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Claudin 4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 86

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Claudin 4 Antibody (32-9400) in WB

          Western blot was performed using Anti- Claudin 4 Monoclonal Antibody (3E2C1) (Product # 32-9400) and a 18 kDa band corresponding to CLDN4 was observed across the positive cells lines tested (MCF7 and HT-29) and not in the negative models (A549 and HeLa) as reported. Membrane enriched extracts (30 µg lysate) of MCF7 (Lane 1), HT-29 (Lane 3), A549 (Lane 2) and HeLa (Lane 4) were electrophoresed using Novex® NuPAGE® 12 % Bis-Tris gel (Product # NP0342BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # IB230... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 4 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 4 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 4 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 4 Antibody in Western Blot, Immunohistochemistry (Paraffin) (WB, IHC (P))
          Claudin 4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 4 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 4 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 4 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Immunohistochemistry (IHC)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Western Blot (WB)
          Claudin 4 Antibody in Immunoprecipitation (IP)
          Claudin 4 Antibody in Peptide array (ARRAY)
          Claudin 4 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Claudin 4 Monoclonal Antibody (3E2C1)

          Product Details

          32-9400

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5 - 3 µg/mL
          View 106 publications 106 publications

          Immunohistochemistry (IHC)

          2-3 µg/mL
          View 51 publications 51 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 37 publications 37 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 11 publications 11 publications

          Immunocytochemistry (ICC/IF)

          -
          View 68 publications 68 publications

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          Peptide Array (Array)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 23 publications 23 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Published species

          Amphibian, Bovine, Dog, Fish, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Sheep, Virus, Xenopus, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3E2C1

          Immunogen

          Synthetic peptide corresponding to a 22 amino acid sequence derived from the C-terminal region of human Claudin-4.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 594 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533096

          Product Specific Information

          Reactivity has been confirmed with human, rat ileum homogenates, and canine MDCK cell lysates by western blotting and immunofluorescence. Reactivity has also been confirmed with formalin-fixed, paraffin-embedded (FFPE) human normal colon and colon cancer tissues by immunohistochemistry.

          For best results in immunohistochemistry with formalin-fixed, paraffin-embedded (FFPE) tissues, heat induced epitope retrieval (HIER) with citrate buffer, pH 6.0, is required prior to staining.

          This antibody reacts specifically with the ~ 22 kDa Claudin-4 protein and does not cross-react with the protein at 55 kDa.

          Target Information

          Claudin-4 belongs to the 20-member family of claudins, transmembrane cellular adhesion proteins localized at epithelial and endothelial tight junctions. Claudins demonstrate distinct, tissue-specific expression patterns, and are essential components of tight junction charge-specific channels that regulate paracellular ion flux. Claudin-4 is a component of tight junction strands and may play a role in internal organ development and function during pre- and postnatal life. The overexpression of Claudin-4 has been shown to decrease paracellular electrical conductance due to a selective decrease in Na+ permeability, with no significant decrease in Cl- permeability. In ovarian carcinoma cells, Claudin-3 and Claudin-4 exhibit upregulation at both mRNA and protein levels. Claudin-4 is also overexpressed in pancreatic cancer tissues and cell lines, and in other epithelial cancers. The Claudin-4 gene is deleted in Williams-Beuren syndrome, a neurodevelopmental disorder affecting multiple systems.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CLAUDIN4; clostridium perfringens enterotoxin receptor; Clostridium perfringens enterotoxin receptor 1; CPE-R; CPE-receptor; h CPE-receptor; unnamed protein product; Williams-Beuren syndrome chromosomal region 8 protein

          View more View less

          Gene Aliases: Cep-r; CPE-R; CPER; CPETR; CPETR1; hCPE-R; WBSCR8

          View more View less

          UniProt ID: (Human) O14493, (Dog) A0A8C0N7E5, (Mouse) O35054

          View more View less

          Entrez Gene ID: (Human) 1364, (Dog) 100856416, (Rat) 304407, (Mouse) 12740

          View more View less

          Function(s)
          transmembrane signaling receptor activity structural molecule activity protein binding identical protein binding tight junction
          Process(es)
          female pregnancy circadian rhythm calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules regulation of cell morphogenesis positive regulation of cell migration response to progesterone establishment of skin barrier renal absorption positive regulation of wound healing paracellular transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch