Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Collagen X Antibodies

          Invitrogen

          Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™

          9 Published Figures
          11 References
          View all (10) Collagen X antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (9)
          Collagen X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Collagen X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Collagen X Antibody (41-9771-82) in IHC (P)

          Immunohistochemistry of formalin-fixed paraffin embedded human infiltrating ductal carcinoma tissue using 5 µg/mL of Mouse IgG1 K Isotype Control eFluor® 570 (left) or 5 µg/mL of Anti-Collagen Type X eFluor® 570 (right). Nuclei are stained with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Collagen X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Collagen X Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed) (IHC, IHC (PFA))
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Immunohistochemistry (IHC)
          Collagen X Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          41-9771-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Guinea pig, Human, Mouse, Pig, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eFluor™ 570, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          X53

          Conjugate

          eFluor™ 570 eFluor™ 570 eFluor™ 570
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          556/569 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2573641

          Product Specific Information

          Description: The monoclonal antibody X53 recognizes human, mouse, porcine, guinea pig, and rat Collagen Type X. X53 does not cross-react with collagen types I, II, II, IV, or VI. Collagen Type X is a homotrimeric short chain collagen. Collagen Type X has limited expression in normal adult tissues, primarily being expressed during bone growth and development. Expression is localized to the peri- and extracellular matrix of hypertrophic chondrocytes during endochondral ossification and also plays a role in cartilage mineralization. Collagen Type X is elevated in many tumor types with expression being localized in the vasculature. Mutations to the gene COL10A1, which encodes human Collagen Type X, is associated with Schmid type metaphyseal chondrodysplasia.

          Applications Reported: This X53 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.

          Applications Tested: This X53 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable.

          Excitation: 555 nm; Emission: 570 nm

          Target Information

          Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: COL10A1var1; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1

          View more View less

          Gene Aliases: Col10; Col10a-1

          View more View less

          UniProt ID: (Human) Q03692, (Mouse) Q05306

          View more View less

          Entrez Gene ID: (Human) 1300, (Pig) 448809, (Dog) 100856711, (Rat) 25681, (Guinea pig) 106026924, (Mouse) 12813

          View more View less

          Function(s)
          protein binding extracellular matrix structural constituent conferring tensile strength extracellular matrix structural protein
          Process(es)
          skeletal system development extracellular matrix organization endochondral ossification cartilage development bone development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline