Product Image
Invitrogen™
eBioscience™ StainTray™
This StainTray™ may be used as a humidified, light-protected chamber for staining slides for immunohistochemistry or immunocytochemistry. The StainTray™ hasRead more
Have Questions?
Catalog NumberQuantity
44-0404-101 each
Catalog number 44-0404-10
Price (USD)
327.18
Special offer
Online exclusive
Ends: 31-Dec-2026
399.00
Save 71.82
Each
Quantity:
1 each
Price (USD)
327.18
Special offer
Online exclusive
Ends: 31-Dec-2026
399.00
Save 71.82
Each
This StainTray™ may be used as a humidified, light-protected chamber for staining slides for immunohistochemistry or immunocytochemistry. The StainTray™ has a black base made of tough ABS plastic withstanding a wide range of chemicals (avoid chlorinated hydrocarbons). It will accept up to 20 slides on four plastic rails covered with a polymer strip to perfectly hold slides even if the tray is held at an angle. When humidity is needed, wells between rails will hold up to 1 mL of water securely without splashing. Middle wells will hold up to 2 mL each. Rails are raised not only to avoid water touching the slides but also to make slides more easily retrievable. The base will also hold excess stain solution dripping from the slides. Four rubber feet ensure greater base stability. Units are stackable for storage and space saving purposes. A black lid is included for work with fluorescent reagents. It is made of ABS and can toleratae a temperature range of -80°C to +80°C.
Dimensions with cover: 38 x 24 x 4.5cm L x W x H (15 x 9 3/8 x 1 3/4 in. L x W xH)
Reported Application
Immunocytochemistry, Immunohistochemical Staining, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Microscopy
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Immunocytochemistry, Immunohistochemical Staining, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Microscopy
Product LineeBioscience
Quantity1 each
Product TypeStain Tray
Slide Capacity20
Unit SizeEach