How can I find a mutant control sample for my Mutation Detection experiment?
You can check the COSMIC database to see if a control sample is identified for your mutation. If one is not available, you can have one synthesized (using our GeneArt Gene Synthesis service, for example).