SulfoLink™ Immobilization Kit for Peptides, 2 mL
SulfoLink™ Immobilization Kit for Peptides, 2 mL
SulfoLink™ Immobilization Kit for Peptides, 2 mL
SulfoLink™ Immobilization Kit for Peptides, 2 mL
Thermo Scientific™

SulfoLink™ Immobilization Kit for Peptides, 2 mL

Thermo Scientific SulfoLink Immobilization Kit for Peptides, 2 mL is a kit containing all the reagents for preparing the peptideRead more
Have Questions?
Catalog NumberQuantity
449995 Column Kit
Catalog number 44999
Price (USD)
512.00
Each
Quantity:
5 Column Kit
Request bulk or custom format
Price (USD)
512.00
Each
Thermo Scientific SulfoLink Immobilization Kit for Peptides, 2 mL is a kit containing all the reagents for preparing the peptide sample for efficient immobilization and five columns containing 2 mL of SulfoLink Resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification.

Features of SulfoLink Immobilization Kit for Peptides, 2 mL:

• Specific to sulfhydryl (-SH) groups—iodoacetyl groups react specifically with sulhydryls to form irreversible thioether bonds
• Fast—spin columns increase protocol speed; prepare and couple peptide samples in 2 hours
• Flexible coupling conditions—use pH 7.5 to 9.0 aqueous buffers, organic solvent (e.g., 20% DMSO) or denaturant (guanidine·HCl), as needed for protein or peptide solubility during coupling reaction
• Easy-to-follow instructions—streamlined protocols for sample preparation, immobilization, and affinity purification
• High capacity—Immobilize 1 to 2 mg of peptide per 2 mL column of SulfoLink Coupling Resin

Applications:
• Immobilize peptides having terminal cysteine residues to purify antibodies that were generated against peptide immunogens prepared by maleimide conjugation
• Immobilize antibodies in oriented manner through hinge-region sulfhydryls to ensure that antigen binding sites are not sterically hindered for antigen affinity purification

When incubated with a solution of peptide that contains reduced cysteine residues, the iodoacetyl groups of SulfoLink Resin react specifically and efficiently with the exposed sulfhydryls (-SH) to form covalent and irreversible thioether bonds that permanently attach the peptide to the resin. The result is a custom-made affinity resin for purification of antibodies, antigens and other molecules of interest.

Related Products
SulfoLink™ Coupling Resin
SulfoLink™ Immobilization Kit for Proteins, 2 mL
SulfoLink™ Immobilization Trial Kit, 2 mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)2 mL
Column TypeAffinity, Agarose Resin
FormatSpin Column
Quantity5 Column Kit
Stationary PhaseHaloacetyl
Sufficient For1 to 2 mg of Sulfhydryl-Peptide per Column
Product LineSulfoLink
TypeImmobilization Kit
Unit SizeEach
Contents & Storage
Sufficient For: Immobilizing 0.1 to 1 mg of sulfhydryl-peptide per column
• SulfoLink Columns, 2 mL, 5 columns
• SulfoLink Coupling Buffer, 120 mL
• Wash Solution, 120 mL
• PBS Pack (makes 500 mL), 1 pack
• Bond-Breaker TCEP Solution, Neutral pH, 0.5 mL
• L-Cysteine·HCl, 100 mg
• Column Accessories

Store at 4°C protected from light.

Frequently asked questions (FAQs)

What is the binding capacity of SulfoLink Resin?

SulfoLink Resin can bind approximately 5 mg of reduced IgG per mL of support. For peptides, the capacity is approximately 1 mg of peptide per mL of gel. Capacity for other compounds will vary.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

When using the SulfoLink Coupling Resins and Kits, my peptide is not soluble in the coupling buffer. What can I do?

SulfoLink Immobilization is compatible with 3-4 M fresh urea or guanidine. Alternatively, dissolve the peptide in 100% DMSO. Add the peptide in DMSO to the coupling buffer so that the DMSO does not exceed 20% of the final solution.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Must I block the resin before each use when using the SulfoLink Coupling Resins and Kits?

No. The resin will no longer react with subsequent sulfhydryls after the ligand is attached and the remaining active sites are blocked.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How many purifications can I perform using the same SulfoLink Column?

The columns can be reused at least 10 times without loss of activity.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How does SulfoLink Immobilization work?

SulfoLink Resin is crosslinked 6% beaded agarose that has been derivatized with a 12-atom spacer arm that ends in an iodoacetyl group. This spacer arm is helpful for peptides or large proteins that might otherwise be sterically hindered in binding their ligand. The iodoacetyl group reacts with free sulfhydryls at pH 7.5-8.5 to form covalent thioether bonds. The sulfhydryl must be present in the reduced form because disulfides will not react with the iodoacetyl group.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.