Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EOMES Antibodies

          Invitrogen

          EOMES Monoclonal Antibody (Dan11mag), PerCP-eFluor™ 710, eBioscience™

          19 Published Figures
          29 References
          View all (42) EOMES antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite EOMES Monoclonal Antibody (Dan11mag), PerCP-eFluor™ 710, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (19)
          EOMES Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          EOMES Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EOMES Antibody (46-4875-82) in Flow

          Surface staining of C57BL/6 splenocytes with Anti-Mouse NK1-1 APC (Product # 17-5941-82), followed by intracellular staining with 0.06 µg of Rat IgG2a K Isotype Control PerCP-eFluor® 710 (Product # 46-4321-82) (left) or 0.06 µg of Anti-Mouse EOMES PerCP-eFluor® 710 (right) using Foxp3/Transcription Factor Buffer Staining Set (Product # 00-5523-00). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)
          EOMES Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          46-4875-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 29 publications 29 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), PerCP-eFluor™ 710, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Dan11mag

          Conjugate

          PerCP-eFluor™ 710 PerCP-eFluor™ 710 PerCP-eFluor™ 710
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 660 
          • APC 
          • eFluor 450
          • eFluor 660
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • Request custom conjugation

          Excitation/Emission Max

          482/708 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10597455

          Product Specific Information

          Description: This Dan11mag antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.

          Applications Reported: This Dan11mag antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This Dan11mag antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol Please see BestProtocols® Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This antibody can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative.

          Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.

          Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: T-box brain protein 2; T-box family member; T-box domain; T-brain-2; unnamed protein product

          View more View less

          Gene Aliases: TBR-2; Tbr2

          View more View less

          UniProt ID: (Mouse) O54839

          View more View less

          Entrez Gene ID: (Mouse) 13813

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding transcription corepressor activity chromatin DNA binding sequence-specific DNA binding RNA polymerase II sequence-specific DNA binding transcription factor binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter endoderm formation mesoderm formation endodermal cell fate specification neuron migration blastocyst development trophectodermal cell differentiation CD8-positive, alpha-beta T cell differentiation involved in immune response chromatin remodeling regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter brain development cardioblast differentiation gene expression regulation of gene expression stem cell population maintenance olfactory bulb development cerebral cortex regionalization forebrain neuron differentiation cerebral cortex neuron differentiation cerebral cortex development neurogenesis cell differentiation neuron differentiation skeletal muscle cell differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter astrocyte differentiation cell differentiation involved in embryonic placenta development mesodermal to mesenchymal transition involved in gastrulation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-nnlvt:80/100.66.74.151:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0