Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PD-L1 (CD274) Antibodies

          Invitrogen

          CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), PerCP-eFluor™ 710, eBioscience™

          46 Published Figures
          41 References
          View all (258) PD-L1 (CD274) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), PerCP-eFluor™ 710, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (46)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 47

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD274 (PD-L1, B7-H1) Antibody (46-5982-80) in Flow

          Staining of C57Bl/6 splenocytes with Anti-Mouse CD3e APC (Product # 17-0031-82) and 0.06 µg of Rat IgG2a K Isotype Control PerCP-eFluor® 710 (Product # 46-4321-82) (left) or 0.06 µg of Anti-Mouse CD274 (B7-H1) PerCP-eFluor® 710 (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          46-5982-80

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 41 publications 41 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, lambda

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), PerCP-eFluor™ 710, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MIH5

          Conjugate

          PerCP-eFluor™ 710 PerCP-eFluor™ 710 PerCP-eFluor™ 710
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 660 
          • APC 
          • Biotin 
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 650
          • Brilliant Violet 786
          • Functional Grade
          • NovaFluor Red 700
          • NovaFluor Red 755
          • NovaFluor Yellow 690
          • NovaFluor Yellow 700
          • PE 
          • PE-Cyanine7
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          482/708 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2573818

          Product Specific Information

          Description: The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody.

          Applications Reported: This MIH5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This MIH5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: B7 homolog 1; PDCD1 ligand 1; pdl-1; sCD274; soluble CD274; soluble PD L1; sPD L1; unnamed protein product

          View more View less

          Gene Aliases: A530045L16Rik; B7h1; Pdcd1l1; Pdcd1lg1; Pdl1

          View more View less

          UniProt ID: (Mouse) Q9EP73

          View more View less

          Entrez Gene ID: (Mouse) 60533

          View more View less

          Function(s)
          transcription coactivator activity protein binding receptor agonist activity
          Process(es)
          negative regulation of T cell mediated immune response to tumor cell immune response signal transduction cell surface receptor signaling pathway positive regulation of cell migration T cell costimulation negative regulation of interferon-gamma production negative regulation of interleukin-10 production positive regulation of interleukin-10 production response to cytokine TRIF-dependent toll-like receptor signaling pathway positive regulation of T cell proliferation negative regulation of T cell proliferation positive regulation of transcription, DNA-templated negative regulation of activated T cell proliferation negative regulation of T cell receptor signaling pathway negative regulation of T cell activation cellular response to lipopolysaccharide negative regulation of tumor necrosis factor superfamily cytokine production positive regulation of activated CD8-positive, alpha-beta T cell apoptotic process negative regulation of CD4-positive, alpha-beta T cell proliferation negative regulation of CD8-positive, alpha-beta T cell activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-x4rxd:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0