Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • B220 (CD45R) Antibodies

          Invitrogen

          CD45R (B220) Monoclonal Antibody (RA3-6B2), APC-eFluor™ 780, eBioscience™

          Advanced Verification
          76 Published Figures
          160 References
          View all (99) B220 (CD45R) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD45R (B220) Monoclonal Antibody (RA3-6B2), APC-eFluor™ 780, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (76)
          • Advanced Verification (1)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 78

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD45R (B220) Antibody (47-0452-82) in Flow

          Staining of C57BL/6 splenocytes with Anti-Mouse CD3e FITC (Product # 11-0031-82) and 0.25 µg of Rat IgG2a kappa Isotype Control APC-eFluor® 780 (Product # 47-4321-82) (left) or 0.25 µg of Anti-Human/Mouse CD45R (B220) APC-eFluor® 780 (right). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          47-0452-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.5 µg/test
          View 159 publications 159 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Bacteria, Fish, Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), APC-eFluor™ 780, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          RA3-6B2

          Conjugate

          APC-eFluor™ 780 APC-eFluor™ 780 APC-eFluor™ 780
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • eFluor 570
          • eFluor 615
          • eFluor 660
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 530
          • NovaFluor Blue 585
          • NovaFluor Blue 610-30S
          • NovaFluor Blue 660-40S
          • NovaFluor Blue 725
          • NovaFluor Blue 745
          • NovaFluor Blue 760
          • NovaFluor Blue 800
          • NovaFluor Red 700
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Violet 745
          • NovaFluor Violet 800
          • NovaFluor Yellow 570
          • NovaFluor Yellow 590
          • NovaFluor Yellow 660
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          756/785 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1518810

          Product Specific Information

          Description: The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220 kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen.

          Applications Reported: This RA3-6B2 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This RA3-6B2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD45R (PTPRC) is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. CD45 is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists in multiple isoforms as a result of alternative splicing, and these isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. Further, these CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. In addition to its role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. Diseases associated with CD45 dysfunction include Severe Combined Immunodeficiency (T-cell negative, B-Cell Negative) and Hepatitis C.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: 200 kDa T-cell leukocyte common antigen isoform; CD45 antigen; CD45; Ly5; glycoprotein 180; glycoprotein isoform Ly-5; leukocyte common antigen; lymphocyte antigen 5; lymphocyte common antigen; protein tyrosine phosphatase; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; T200 glycoprotein; T200 leukocyte common antigen; unnamed protein product

          View more View less

          Gene Aliases: B220; CD45; CD45R; GP180; IMD105; L-CA; LCA; loc; Ly-5; LY5; Lyt-4; T200

          View more View less

          UniProt ID: (Human) P08575, (Mouse) P06800

          View more View less

          Entrez Gene ID: (Human) 5788, (Mouse) 19264

          View more View less

          Function(s)
          protein tyrosine phosphatase activity transmembrane receptor protein tyrosine phosphatase activity receptor binding protein binding heparin binding protein kinase binding protein tyrosine kinase inhibitor activity ankyrin binding spectrin binding heparan sulfate proteoglycan binding identical protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter natural killer cell differentiation negative regulation of T cell mediated cytotoxicity positive regulation of T cell mediated cytotoxicity negative regulation of cytokine-mediated signaling pathway hematopoietic progenitor cell differentiation positive regulation of immunoglobulin production positive regulation of T cell mediated immunity regulation of humoral immune response mediated by circulating immunoglobulin positive regulation of humoral immune response mediated by circulating immunoglobulin negative regulation of protein kinase activity protein dephosphorylation negative regulation of cell adhesion involved in substrate-bound cell migration leukocyte cell-cell adhesion signal transduction response to gamma radiation regulation of gene expression dephosphorylation B cell differentiation T cell differentiation positive regulation of B cell proliferation negative regulation of protein autophosphorylation regulation of interleukin-8 production negative regulation of interleukin-2 production positive regulation of interleukin-2 production positive regulation of tumor necrosis factor production heterotypic cell-cell adhesion T cell proliferation B cell proliferation positive regulation of T cell proliferation positive regulation of MAP kinase activity positive regulation of MAPK cascade cell cycle phase transition plasma membrane raft distribution positive thymic T cell selection negative thymic T cell selection positive regulation of T cell differentiation positive regulation of gamma-delta T cell differentiation negative regulation of JAK-STAT cascade positive regulation of alpha-beta T cell proliferation positive regulation of isotype switching to IgG isotypes bone marrow development stem cell development negative regulation of peptidyl-tyrosine phosphorylation regulation of phagocytosis positive regulation of phagocytosis positive regulation of calcium-mediated signaling T cell receptor signaling pathway B cell receptor signaling pathway regulation of T cell receptor signaling pathway positive regulation of antigen receptor-mediated signaling pathway release of sequestered calcium ion into cytosol defense response to virus regulation of cell cycle positive regulation of Fc receptor mediated stimulatory signaling pathway negative regulation of ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade negative regulation of interleukin-4-mediated signaling pathway positive regulation of hematopoietic stem cell migration positive regulation of stem cell proliferation regulation of extrinsic apoptotic signaling pathway positive regulation of extrinsic apoptotic signaling pathway cell surface receptor signaling pathway T cell activation positive regulation of protein kinase activity negative regulation of microglial cell activation response to aldosterone DN2 thymocyte differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline