Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CSF1R (CD115) Antibodies

          Invitrogen

          CD115 (c-fms) Monoclonal Antibody (AFS98), APC-eFluor™ 780, eBioscience™

          27 Published Figures
          28 References
          View all (90) CSF1R (CD115) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD115 (c-fms) Monoclonal Antibody (AFS98), APC-eFluor™ 780, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (27)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 28

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD115 (c-fms) Antibody (47-1152-82) in Flow

          Mouse bone marrow cells were cultured for 3 days with Mouse M-CSF Recombinant Protein (Product # 14-8983-80). Cells were stained with Anti-Mouse CD11b PE (Product # 12-0112-82) and 0.125 µg of Rat IgG2a K Isotype Control APC-eFluor® 780 (Product # 47-4321-82) (left) or 0.125 µg of Anti-Mouse CD115 (c-fms) APC-eFluor® 780 (right). Total viable cells, as determined by Fixable Viability Dye eFluor® 506 (Product # 65-0866-14), were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          47-1152-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 28 publications 28 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Fish, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), APC-eFluor™ 780, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AFS98

          Conjugate

          APC-eFluor™ 780 APC-eFluor™ 780 APC-eFluor™ 780
          • Unconjugated
          • Alexa Fluor 488 
          • APC 
          • Biotin 
          • Brilliant UV 563
          • Functional Grade
          • NovaFluor Red 685
          • NovaFluor Red 710
          • NovaFluor Yellow 610
          • NovaFluor Yellow 730
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          756/785 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2573970

          Product Specific Information

          Description: The AFS98 monoclonal antibody reacts with the mouse CD115 molecule, a receptor for macrophage colony stimulating factor (M-CSF) or colony stimulating factor-1 (CSF-1). CD115 is expressed by monocyte, macrophage, osteoclast, and some epithelial cells. It is a 150 kDa c-fms gene product and belongs to immunoglobulin family. CSF-1 signaling through CSF-1R regulates the proliferation and differentiation of cells in the monocytic lineage.

          Applications Reported: This AFS98 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This AFS98 antibody has been tested by flow cytometric analysis of mouse bone marrow-derived macrophages. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          APC-eFluor® 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: c-fms; CSF-1 receptor; CSF-1-R; CSF-R1; CSFIR; EC 2.7.10.1; kinase CSFR; OTTHUMP00000224093; proto-oncogene c-Fms; proto-oncogene fms; unnamed protein product

          View more View less

          Gene Aliases: CD115; CSF-1R; Csfmr; Fim-2; Fim2; Fms; M-CSF-R; M-CSFR

          View more View less

          UniProt ID: (Mouse) P09581

          View more View less

          Entrez Gene ID: (Mouse) 12978

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity macrophage colony-stimulating factor receptor activity protein binding ATP binding growth factor binding protein phosphatase binding cytokine binding protein homodimerization activity
          Process(es)
          microglial cell activation neutrophil homeostasis positive regulation of protein phosphorylation myeloid leukocyte differentiation response to ischemia response to stress signal transduction transmembrane receptor protein tyrosine kinase signaling pathway axon guidance cell proliferation positive regulation of cell proliferation negative regulation of cell proliferation regulation of cell shape anatomical structure morphogenesis positive regulation of gene expression positive regulation of macrophage chemotaxis negative regulation of myotube differentiation microglia development cell migration peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway telencephalon development dentate gyrus development olfactory bulb development forebrain neuron differentiation regulation of cell morphogenesis hemopoiesis osteoclast differentiation positive regulation of cell migration positive regulation of bone mineralization negative regulation of epithelial cell differentiation ruffle organization positive regulation of chemokine production positive regulation of tumor necrosis factor production regulation of mononuclear cell proliferation regulation of actin cytoskeleton organization monocyte homeostasis cellular response to macrophage colony-stimulating factor stimulus macrophage colony-stimulating factor signaling pathway positive regulation of tyrosine phosphorylation of STAT protein negative regulation of apoptotic process regulation of MAPK cascade positive regulation by host of viral process regulation of bone resorption cell-cell junction maintenance positive regulation of osteoclast differentiation protein autophosphorylation positive regulation of lymphocyte proliferation positive regulation of protein kinase B signaling positive regulation of protein tyrosine kinase activity microglial cell proliferation positive regulation of astrocyte activation positive regulation of ERK1 and ERK2 cascade positive regulation of leukocyte proliferation cellular response to cytokine stimulus positive regulation of macrophage proliferation positive regulation of cell cycle G1/S phase transition negative regulation of platelet formation regulation of macrophage migration positive regulation of macrophage migration neuron projection extension positive regulation of cell motility positive regulation of myoblast proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch