Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Actin Muscle Antibodies

          Invitrogen

          Actin (muscle) Monoclonal Antibody (HHF35), Alexa Fluor™ 488, eBioscience™

          1 Published Figure
          5 References
          View all (19) Actin Muscle antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Actin (muscle) Monoclonal Antibody (HHF35), Alexa Fluor™ 488, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          Actin (muscle) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Actin (muscle) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Actin (muscle) Antibody (53-6496-80) in ICC/IF

          Immunocytochemistry on fixed and permeabilized C2C12 cells stained with 30 µg/mL Mouse IgG1 Alexa Fluor® 488 Isotype Control (left) or 30 µg/mL Anti-Actin (muscle) Alexa Fluor® 488 (right). Nuclei are stained with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Actin (muscle) Antibody in Immunocytochemistry (ICC/IF)
          Actin (muscle) Antibody in Immunohistochemistry (IHC)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          53-6496-80

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          30 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Horse, Guinea pig, Human, Mouse, Non-human primate, Pig, Rat

          Published species

          Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Alexa Fluor™ 488, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HHF35

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2574432

          Product Specific Information

          Description: The monoclonal antibody HHF35 recognizes muscle actin; specifically the alpha and gamma form found in skeletal cardiac, and smooth muscle. No reactivity to the beta form is observed. The 42 kDa protein is a main component of the cytoskeletal structural network. The HHF35 antibody has been shown to recognize actin from smooth muscle cells of the heart, bladder, uterus, gastrointestinal tract, and diaphragm. The antibody stains tumors of muscle origin such as leiomyomas, leiosarcomes and rhabdomyosacromas.

          The HHF35 antibody recognizes human, mouse, rat, bovine, canine and non-human primate actin.

          Applications Reported: This HHF35 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.

          Applications Tested: This HHF35 antibody has been tested by immunocytochemistry and can be used at less than or equal to 30 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ACTA1; ACTA2; ACTB; ACTC; ACTG2; actin; actin alpha 1; actin alpha cardiac; actin homolog; actin, alpha 1, skeletal muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin-like protein; alfa sma; alpha actin; alpha actin 1; alpha actin 2; alpha smooth muscle actin; alpha-actin; alpha-actin cardiac; alpha-actin-1; alpha-actin-2; alpha-actin-3; alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; aSMA; aSmooth Muscle Actin; B-actin; beta actin; beta-actin FE-3; cardiac actin; cardiac alpha actin; cardiac muscle alpha actin 1; cell growth-inhibiting gene 46 protein; cytoplasmic actin; cytoskeletal gamma-actin; epididymis luminal protein 176; gamma-2-actin; gamma-actin; gamma-enteric smooth muscle actin; GIG46; nemaline myopathy type 3; RP5-1068B5.2; skeletal alpha actin; skeletal muscle alpha-actin; skeletal muscle alpha-actin 1; smooth muscle alpha-actin; smooth muscle gamma-actin; unnamed protein product; vascular smooth muscle alpha-actin

          View more View less

          Gene Aliases: 0610041G09Rik; a-SMA; ACT; ACT-4; Act4; ACTA; Acta-2; ACTA1; ACTA3; ACTC; Actc-1; ACTE; ACTG; ACTGE; actin; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; alphaSMA; ASD5; ASMA; CFTD; CFTD1; CFTDM; CMD1R; CMH11; CMYO2A; CMYO2B; CMYO2C; CMYP2A; CMYP2B; CMYP2C; DFNA20; DFNA26; E51; HEL-176; LVNC4; MMIHS5; MPFD; NEM1; NEM2; NEM3; SHPM; SMAalpha; SMalphaA; SMDYS; SMGA; VSCM; VSCM1

          View more View less

          UniProt ID: (Human) P68133, (Bovine) P68138, (Pig) P68137, (Bovine) P62739, (Human) P62736, (Bovine) Q3ZC07, (Human) P68032, (Human) P63261, (Bovine) P63258, (Bovine) Q5E9B5, (Human) P63267, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Rat) P68035, (Mouse) P68033, (Mouse) P63260, (Rat) P63259, (Rat) P63269, (Mouse) P63268

          View more View less

          Entrez Gene ID: (Horse) 100065454, (Human) 58, (Bovine) 281592, (Pig) 100154254, (Dog) 488984, (Pig) 733615, (Dog) 477587, (Bovine) 515610, (Horse) 100062175, (Human) 59, (Dog) 478250, (Bovine) 533219, (Horse) 100057678, (Human) 70, (Pig) 100152267, (Dog) 475923, (Human) 71, (Bovine) 404122, (Pig) 397653, (Horse) 100050139, (Bovine) 281595, (Pig) 100520667, (Human) 72, (Horse) 100057561, (Dog) 475792, (Mouse) 11459, (Guinea pig) 100716933, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Guinea pig) 100722917, (Rat) 29275, (Guinea pig) 100731953, (Mouse) 11464, (Mouse) 11465, (Rat) 287876, (Guinea pig) 100731713, (Rat) 25365, (Guinea pig) 100716255, (Mouse) 11468

          View more View less

          Function(s)
          structural constituent of cytoskeleton protein binding ATP binding ATPase activity myosin binding ADP binding protein kinase binding microfilament motor activity profilin binding ubiquitin protein ligase binding identical protein binding structural constituent of postsynaptic actin cytoskeleton actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          muscle contraction positive regulation of gene expression skeletal muscle thin filament assembly skeletal muscle fiber development mesenchyme migration regulation of blood pressure response to virus vascular smooth muscle contraction glomerular mesangial cell development cytoskeleton organization positive regulation of hepatic stellate cell migration positive regulation of hepatic stellate cell contraction positive regulation of ERK1 and ERK2 cascade cellular response to transforming growth factor beta stimulus juxtaglomerular apparatus development positive regulation of hepatic stellate cell activation actin filament organization actin filament-based movement actomyosin structure organization actin-myosin filament sliding cardiac myofibril assembly cardiac muscle tissue morphogenesis heart contraction angiogenesis morphogenesis of a polarized epithelium axonogenesis positive regulation of cell migration maintenance of blood-brain barrier cell motility regulation of stress fiber assembly regulation of focal adhesion assembly platelet aggregation positive regulation of wound healing postsynaptic actin cytoskeleton organization tight junction assembly regulation of transepithelial transport regulation of synaptic vesicle endocytosis protein localization to bicellular tight junction response to ethanol cellular response to interleukin-6 cellular response to acetaldehyde response to mechanical stimulus cellular response to potassium ion skeletal muscle fiber adaptation response to steroid hormone response to xenobiotic stimulus negative regulation of apoptotic process cardiac muscle contraction cytoplasmic actin-based contraction involved in cell motility sarcomere organization cellular response to interferon-gamma response to calcium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0