Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ROR gamma (t) Antibodies

          Invitrogen

          ROR gamma (t) Monoclonal Antibody (AFKJS-9), PE-eFluor™ 610, eBioscience™

          26 Published Figures
          19 References
          View all (14) ROR gamma (t) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ROR gamma (t) Monoclonal Antibody (AFKJS-9), PE-eFluor™ 610, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (26)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 27

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ROR gamma (t) Antibody (61-6988-80) in Flow

          C57BL/6 mouse thymocytes were surface-stained with CD8a Monoclonal Antibody, eFluor 450 (Product # 48-0081-82) and CD4 Monoclonal Antibody, FITC (Product # 11-0041-82). Cells were then stained intracellularly, using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol, with 0.5 µg of Rat IgG2a kappa Isotype Control, PE-eFluor 610 (Product # 61-4321-82) (left) or 0.5 µg of ROR gamma (t) Monoclonal Antibody, PE-eFluor 610 (right). CD4+CD8+ double-positive (purple histogram) or CD4+CD8- single-positive (blue ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Immunohistochemistry (IHC)
          ROR gamma (t) Antibody in Western Blot (WB)
          ROR gamma (t) Antibody in Western Blot (WB)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)
          ROR gamma (t) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          61-6988-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          1.0 µg/test
          View 16 publications 16 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rhesus monkey

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), PE-eFluor™ 610, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AFKJS-9

          Conjugate

          PE-eFluor™ 610 PE-eFluor™ 610 PE-eFluor™ 610
          • Unconjugated
          • APC 
          • PE 
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          565/606 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2848499

          Product Specific Information

          Description: The AFKJS-9 monoclonal antibody reacts with the mouse, human and rhesus monkey ROR gamma (t) protein. ROR gamma is a member of the retinoic acid-related orphan receptor (ROR) family, which also includes ROR alpha and ROR beta. ROR family proteins are ligand-dependent transcription factors that play roles in multiple physiological processes. ROR gamma is expressed in several tissues including liver, lung, muscle, heart and kidney. Furthermore, it was discovered that alternative transcription results in the expression of an isoform, ROR gamma (t), which is expressed exclusively in cells of the lymphoid compartment, namely CD4+CD8+ "double-positive" thymocytes, Th17 cells of the periphery and lymphoid tissue inducer (LTi) cells of lymphoid organs.

          The ROR gamma (t) isoform differs from ROR gamma by three unique amino acids at its amino terminus. Therefore, the AFKJS-9 antibody will react with both the ROR gamma and ROR gamma (t) isoforms.

          Applications Reported: This AFKJS-9 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This AFKJS-9 antibody has been tested by intracellular staining followed by flow cytometric analysis of mouse thymocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to "Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins" located at Flow Protocols . This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          PE-eFluor 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red). Please make sure that your instrument is capable of detecting this fluorochrome.

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Target Information

          ROR gamma, a NR1 Thyroid Hormone-Like receptor, has been shown to affect thymopoiesis, bone metabolism, T-cell apoptosis, and lymphoid organogenesis. ROR gamma has been shown to promote thymocyte survival by activating the expression of the antiapoptotic protein Bcl-x(L). ROR gamma is also required for the development of lymph nodes and Peyer's patches. It has been shown that ROR gamma(t), a thymus-specific isoform of ROR gamma from mouse, inhibits Fas ligand expression and cytokine secretion in immature thymocytes. ROR gamma binds as a monomer to response elements composed of a single core motif, GGTCA, preceded by a 6 bp AT-rich sequence. ROR gamma expression has been documented in mouse thymus, adipose, bone, skeletal muscle, liver, and kidney, and in human skeletal muscle. ESTs have been isolated from human tissue libraries, including cancerous head/neck, germ cell, and prostate, and normal colon, head/neck, lung, liver/spleen, and pancreas.

          ROR gammat t (RORgt) is an isoform of RORC.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: MGC129539; nuclear receptor RZR-gamma; nuclear receptor subfamily 1 group F member 3; orphan nuclear receptor; RAR-related orphan receptor C; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; ROR Gamma; ROR gamma t; RORC; RORgt; RORyt; RP11-98D18.11-001; thymus orphan receptor; unnamed protein product

          View more View less

          Gene Aliases: EGK_01272; IMD42; NR1F3; RORG; RORgamma; RZR-GAMMA; RZRG; Thor; TOR

          View more View less

          UniProt ID: (Human) P51449, (Mouse) P51450

          View more View less

          Entrez Gene ID: (Rhesus monkey) 717052, (Human) 6097, (Mouse) 19885

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding oxysterol binding zinc ion binding sequence-specific DNA binding transcription factor activity, direct ligand regulated sequence-specific DNA binding sequence-specific double-stranded DNA binding transcription coactivator binding C4 zinc finger nuclear receptor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter tolerance induction in gut-associated lymphoid tissue regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter xenobiotic metabolic process regulation of gene expression regulation of glucose metabolic process regulation of steroid metabolic process intracellular receptor signaling pathway circadian regulation of gene expression cellular response to sterol T-helper cell differentiation positive regulation of circadian rhythm regulatory T cell differentiation regulation of fat cell differentiation positive regulation of transcription, DNA-templated lymph node development Peyer's patch development adipose tissue development negative regulation of thymocyte apoptotic process T-helper 17 cell differentiation protein phosphorylation cell differentiation T cell differentiation in thymus regulation of gamma-delta T cell differentiation alpha-beta T cell differentiation mucosal-associated lymphoid tissue development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0