Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PD-L1 (CD274) Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.

          Invitrogen

          CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), Super Bright™ 780, eBioscience™

          48 Published Figures
          39 References
          View all (281) PD-L1 (CD274) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols

          Cite CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), Super Bright™ 780, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (48)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 49

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD274 (PD-L1, B7-H1) Antibody (78-5982-80) in Flow

          Staining of BALB/c splenocytes with CD3e Monoclonal Antibody, FITC (Product # 11-0031) and 0.03 µg of Rat IgG2a kappa Isotype Control, Super Bright 780 (Product # 78-4321) (left) or 0.03 µg of CD274 Monoclonal Antibody, Super Bright 780 (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Functional Assay (Functional)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          78-5982-80

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          0.06 µg/test
          View 28 publications 28 publications

          Neutralization (Neu)

          -
          View 3 publications 3 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, lambda

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Super Bright™ 780, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MIH5

          Conjugate

          Super Bright™ 780 Super Bright™ 780 Super Bright™ 780
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 660 
          • APC 
          • Biotin 
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 650
          • Brilliant Violet 786
          • Functional Grade
          • NovaFluor Red 700
          • NovaFluor Red 755
          • NovaFluor Yellow 690
          • NovaFluor Yellow 700
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          413/780 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_2724080

          Product Specific Information

          Description: The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody.

          Applications Reported: This MIH5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This MIH5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Super Bright 780 can be excited with the violet laser line (405 nm) and emits at 780 nm. We recommend using a 780/60 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.

          When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

          In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product # 01-2222-42) as compared to single-color stained cells. In such circumstances, we would recommend setting compensation with cells. We have also observed this in some experiments using AbC Total Antibody Compensation beads (Product # A10497).

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 405 nm; Emission: 780 nm; Laser: Violet Laser

          Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

          Target Information

          Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch