AEBSF Protease Inhibitor
Thermo Scientific™

AEBSF Protease Inhibitor

Thermo Scientific AEBSF Protease Inhibitor irreversably inhibits serine proteases. Thermo Scientific Protease Inhibitors are small packages of individual protease inhibitorRead more
Have Questions?
Catalog NumberQuantity
78431100 mg
Catalog number 78431
Price (USD)
165.65
Online exclusive
182.00
Save 16.35
Each
Quantity:
100 mg
Price (USD)
165.65
Online exclusive
182.00
Save 16.35
Each
Thermo Scientific AEBSF Protease Inhibitor irreversably inhibits serine proteases. Thermo Scientific Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.

Features of Thermo Scientific AEBSF Protease Inhibitor:

• High-quality reagents in convenient and affordable package sizes
• Purchase individually to prepare customized protease inhibitor cocktails
• Supplement Halt Protease Inhibitor Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents

The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A and PMSF. Although Thermo Scientific Halt Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail or combine several components to make customized protease inhibitor cocktails for specialized applications.

Related Products
Pierce™ Protease and Phosphatase Inhibitor Mini Tablets, EDTA Free
Pierce™ Protease Inhibitor Mini Tablets, EDTA Free
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce AEBSF Protease Inhibitor
InhibitorsProtease Inhibitor
Quantity100 mg
Reagent TypeProtease Inhibitor
FormLyophilized
Product LinePierce
Product TypeProtease Inhibitor
Unit SizeEach
Contents & Storage
Store between 2°C and 25°C

Frequently asked questions (FAQs)

What formats of protease inhibitors do you offer?

We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Do you offer any of the components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets individually?

Yes, the following components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are offered separately:

- AEBSF (Cat. No. 78431)
- Aprotinin (Cat. No. 78432)
- Bestatin (Cat. No. 8433)
- E-64 (Cat. No. 78434)
- Leupeptin (Cat. No. 78435)
- Pepstatin A (Cat. No. 78436)
- EDTA (Cat. No. 17892).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.