Product Image
Applied Biosystems™

GeneTitan™ Hybridization, Wash, and Stain Kit for 3' IVT Arrays

The GeneTitan™ Hybridization, Wash, and Stain Kit for 3'-IVT Arrays contains pre-formulated solutions to process 3'-IVT array plates on theLeia mais
Have Questions?
Número do catálogoQuantity
9015301 kit
Número do catálogo 901530
Preço (BRL)
-
Quantity:
1 kit
The GeneTitan™ Hybridization, Wash, and Stain Kit for 3'-IVT Arrays contains pre-formulated solutions to process 3'-IVT array plates on the GeneTitan™ System. The reagents are formulated into the fewest number of individual components possible, reducing preparation and handling steps. With enhanced performance, fewer required reagents, and a simple protocol, target preparation has never been so easy.

The GeneTitan Hybridization, Wash, and Stain Kit for 3'-IVT Arrays includes two boxes:

The first box contains sufficient amounts of the hybridization and stain components to process one 96-array plate, four 24-array plates, or six 16-array plates. The hybridization components include the solutions required for preparation of the hybridization cocktail containing the target. The stain components include Stain Cocktail 1 and 3, Stain Cocktail 2, and the Array Holding Buffer. These solutions are used during the processing of the arrays in the GeneTitan System.

The second box includes Wash Buffer A and Wash Buffer B, also required for the processing of the arrays. There is sufficient Wash Buffer A and B to process one plate in any format. For each additional plate that is processed, an additional Wash Buffer A and B module must be purchased.

The Stain Cocktail 1 and 3, Stain Cocktail 2, Array Holding Buffer, and Wash Buffers A and B are ready-to-use solutions. The hybridization cocktail is prepared by combining the Hybridization Mix and nuclease-free water with the target prepared for the specific target labeling assay (IVT or WT).

The GeneTitan Hybridization, Wash, and Stain Kit for 3'-IVT Arrays should always be stored at 2 to 8° C.

Related Links
B2 Control Oligo (3nM)
GeneTitan™ Hybridization, Wash, and Stain Kit for WT Array Plates
GeneTitan™ Wash Buffers A and B Module
GeneChip™ Hybridization Control Kit
Genetitan™ miRNA Array Holding Buffer
For Research Use Only. Not for use in diagnostic procedures.
Especificações
For Use With (Application)Transcriptome Profiling
Quantity1 kit
Reagent TypeHybridization, Wash, Stain
TypeHybridization, Wash, Stain Kit
Format24-array Plate, 96-array Plate
Unit SizeEach