Product Image
Applied Biosystems™
GeneTitan™ Wash Buffers A and B Module
The Genetitan™ Wash Buffer A and Wash Buffer B Module is required for processing Applied Biosystems™ 3' IVT, miRNA profilingMás información
Have Questions?
Número de catálogo 901583
Precio (USD)
-
The Genetitan™ Wash Buffer A and Wash Buffer B Module is required for processing Applied Biosystems™ 3' IVT, miRNA profiling plates, and whole-transcript (WT) array plates on the GeneTitan System. There is sufficient Wash Buffer A and B to process one plate in any format. For each additional plate that is processed, an additional Wash Buffer A and B module must be purchased.
Related Links
GeneTitan™ Hybridization, Wash, and Stain Kit for WT Array Plates
GeneTitan™ Hybridization, Wash, and Stain Kit for 3' IVT Arrays
GeneTitan™ Wash Buffers A and B Module
GeneChip™ Hybridization Control Kit
B2 Control Oligo (3nM)
GeneChip™ miRNA 4.1 24-Array Plate and Trays
GeneChip™ miRNA 4.1 96-Array Plate and Trays
GeneTitan™ miRNA Array Holding Buffer
GeneChip™ miRNA 3.1 24-Array Plate and Trays
GeneChip™ miRNA 3.1 96-Array Plate and Trays
FlashTag™ Biotin HSR RNA Labeling Kits
FlashTag™ Biotin HSR RNA Labeling Kits
GeneTitan™ Hybridization Module for miRNA Plates
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Nombre del producto químico o materialWash Buffer
Para utilizar con (aplicación)miRNA Profiling
Cantidad1 kit
Tipo de soluciónWash Buffer
Unit SizeEach
Preguntas frecuentes
What signal and fold-change concordance can I expect between Affymetrix miRNA 4.0 and miRNA 4.1 Arrays and miRNA 3.0 and miRNA 3.1 Arrays?
You should expect high signal and fold-change concordance for transcripts with identical probes on the miRNA 4.0 and miRNA 4.1 Arrays and the miRNA 3.0 and miRNA 3.1 Arrays. The snoRNA, scaRNA, and precursor miRNA probe sets were reselected, and for some probe sets in these categories, the probes are not identical between the two array designs. Because the probes selected may be different, one may observe changes in the probe set signal summary for a given target. This is possible as different probe sequences will have varying affinity to the target and may show a higher or lower absolute probe signal depending on the GC content of the probes. However, fold changes are typically well-correlated to the legacy probe set unless there is cross-hybridization in the legacy probe set. Our expectation is that more unique and specific probes result in more specific hybridization to the intended target, and therefore a better performing probe set. Target preparation replicates of human brain and lung samples were pooled and hybridized to four miRNA 3.0 Arrays and four miRNA 4.0 Arrays. The Pearson product moment correlation was calculated from the median signal of matched probe sets and from the median fold change of detected matched probe sets on the miRNA 3.0 Array compared to the miRNA 4.0 Array. Probe sets were defined as detected if the median DABG p-value of the four miRNA 3.0 Array brain replicates was less than 0.06. Two subsets of matched probe sets were used for miRNA 3.0 Array to miRNA 4.0 Array signal and fold change correlation: All human probe sets including those that do not necessarily share identical probe sequences and human probe sets for which the probe sequences are identical on both the miRNA 3.0 and miRNA 4.0 Arrays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Do we have to use any Affymetrix Reagents with our assay?
GeneChip Hybridization Control Kit 30 reactions
GeneTitan Hybridization, Wash, and Stain Kit for miRNA Array Plates 96 reactions
GeneTitan Hybridization Module for miRNA Plates 96 reactions
Affymetrix FlashTag Biotin HSR RNA Labeling Kit 10 reactions
Affymetrix FlashTag Biotin HSR RNA Labeling Kit 30 reactions
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What kind of dynamic range can I expect from Affymetrix miRNA Array Plates?
Affymetrix miRNA Array Plate provides a similar dynamic range to GeneChip miRNA Array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do results from GeneChip miRNA Cartridge Array compare to Affymetrix miRNA Array Plates?
When evaluating non-control probe sets in common to both arrays, we observed very high signal and fold-change correlation for GeneChip miRNA Cartridge Arrays compared to Affymetrix miRNA Array Plates (Pearson correlation coefficients of 0.97-0.98 for signal correlation and Pearson correlation coefficients of 0.96-0.98 for fold-change correlation).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the tolerance around hybridization time when working with Affymetrix miRNA Array strips?
We recommend maintaining consistency in array processing protocols to minimize introduction of variability. Our recommended hybridization time for Thermo Fisher Scientific miRNA Array Strip is 20 hrs.
Thermo Fisher Scientific miRNA Array Strip should be hybridized no less than 20 hrs and no longer than 24 hrs. We strongly encourage maintaining consistent hybridization times to avoid false positives with respect to differential expression due to hybridization time.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.